Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27469196-27469465 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA2
Entrez Gene ID
1135
Gene Name
cholinergic receptor nicotinic alpha 2 subunit
Gene Aliases
-
UniGene
Hs.57718
Gene Chromosome Location
Chr.8: 27459761-27479296 on Build GRCh38
Transcripts
NM_000742.3 NM_001282455.1 XM_005273397.2 XM_006716282.1 XM_011544388.1 XM_011544389.1 XM_017013003.1 XM_017013004.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4436455 c.324C>T p.N108N Chr.8: 27469350-27469350 on Build GRCh38
4600299 c.320C>G p.T107S Chr.8: 27469354-27469354 on Build GRCh38
4453506 c.304A>C p.N102H Chr.8: 27469370-27469370 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768490793 Chr.8: 27469327-27469327 on Build GRCh38
rs749863419 Chr.8: 27469297-27469297 on Build GRCh38
rs139680977 Chr.8: 27469322-27469323 on Build GRCh38
rs774451576 Chr.8: 27469418-27469418 on Build GRCh38
rs1003641424 Chr.8: 27469224-27469224 on Build GRCh38
rs796052293 Chr.8: 27469326-27469327 on Build GRCh38
rs757951570 Chr.8: 27469303-27469303 on Build GRCh38
rs1002474650 Chr.8: 27469441-27469441 on Build GRCh38
rs913563840 Chr.8: 27469352-27469352 on Build GRCh38
rs1034654767 Chr.8: 27469439-27469439 on Build GRCh38
rs796052295 Chr.8: 27469373-27469373 on Build GRCh38
rs944982830 Chr.8: 27469384-27469384 on Build GRCh38
rs369707163 Chr.8: 27469391-27469391 on Build GRCh38
rs1046858418 Chr.8: 27469420-27469420 on Build GRCh38
rs770131239 Chr.8: 27469341-27469341 on Build GRCh38
rs951972094 Chr.8: 27469235-27469235 on Build GRCh38
rs544939077 Chr.8: 27469350-27469350 on Build GRCh38
rs746630071 Chr.8: 27469325-27469325 on Build GRCh38
rs910397272 Chr.8: 27469312-27469312 on Build GRCh38
rs968216005 Chr.8: 27469258-27469258 on Build GRCh38
rs1009611030 Chr.8: 27469230-27469230 on Build GRCh38
rs926906905 Chr.8: 27469288-27469288 on Build GRCh38
rs796052296 Chr.8: 27469367-27469367 on Build GRCh38
rs776047724 Chr.8: 27469385-27469386 on Build GRCh38
rs745364666 Chr.8: 27469387-27469387 on Build GRCh38
rs757075915 Chr.8: 27469294-27469294 on Build GRCh38
rs200926431 Chr.8: 27469296-27469296 on Build GRCh38
rs748560508 Chr.8: 27469336-27469336 on Build GRCh38
rs140159249 Chr.8: 27469343-27469343 on Build GRCh38
rs564532686 Chr.8: 27469351-27469351 on Build GRCh38
rs770464377 Chr.8: 27469321-27469322 on Build GRCh38
rs985900885 Chr.8: 27469283-27469283 on Build GRCh38
rs747401096 Chr.8: 27469390-27469390 on Build GRCh38
rs756788513 Chr.8: 27469282-27469282 on Build GRCh38
rs886044444 Chr.8: 27469353-27469353 on Build GRCh38
rs559195609 Chr.8: 27469233-27469233 on Build GRCh38
rs779763586 Chr.8: 27469304-27469304 on Build GRCh38

View Full Product Details