Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008895.2 | NP_001008895.1 | 8 - 9 | 971 | 76 |
NM_001278513.1 | NP_001265442.1 | 8 - 9 | 1115 | 76 | |
NM_001278514.1 | NP_001265443.1 | 8 - 9 | 783 | 76 | |
NM_003589.2 | NP_003580.1 | 8 - 9 | 797 | 76 | |
XM_011537523.2 | XP_011535825.1 | 7 - 8 | 1331 | 76 | |
GenBank mRNA | AB012193.1 | - | 2 - 3 | 145 | 76 |
AF077188.1 | - | 8 - 9 | 708 | 76 | |
AK314835.1 | - | 8 - 9 | 1070 | 76 | |
AL833355.1 | - | 7 - 8 | 735 | 76 | |
AY365124.1 | - | 8 - 9 | 848 | 76 | |
BC008308.1 | - | 8 - 9 | 859 | 76 |