Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_197975.2 | NP_932079.1 | 2 - 3 | 581 | 86 |
GenBank mRNA | AB020625.1 | - | 2 - 3 | 555 | 86 |
AK025267.1 | - | 2 - 3 | 559 | 86 | |
AY358385.1 | - | 2 - 3 | 530 | 86 | |
BC100800.1 | - | 2 - 3 | 467 | 86 | |
BC100801.1 | - | 2 - 3 | 467 | 86 | |
BC100802.1 | - | 2 - 3 | 467 | 86 |