Assay Design



Gene Transcripts

Gene Symbol
TET2
Entrez Gene ID
54790
Gene Name
tet methylcytosine dioxygenase 2
Gene Aliases
KIAA1546, MDS
Chromosome Location
Chr.4: 105145875 - 105279803 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127208.2 NP_001120680.1 2 - 3 436 62
NM_017628.4 NP_060098.3 2 - 3 353 62
XM_006714242.3 XP_006714305.1 2 - 3 280 62
XM_017008319.1 XP_016863808.1 2 - 3 280 62
GenBank mRNA AI084057.1 - 2 - 3 335 62
AK055149.1 - 2 - 3 243 62
BX640738.1 - 2 - 3 354 62
DA391215.1 - 2 - 3 245 62

View Full Product Details