Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 13564023-13564270 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIN2B
Entrez Gene ID
2904
Gene Name
glutamate ionotropic receptor NMDA type subunit 2B
Gene Aliases
EIEE27, GluN2B, MRD6, NMDAR2B, NR2B, hNR3
UniGene
Hs.504844
Gene Chromosome Location
Chr.12: 13537337-13982012 on Build GRCh38
Transcripts
NM_000834.3 XM_005253351.3 XM_011520628.2 XM_011520629.2 XM_017019219.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5469937 c.3202G>A p.V1068I Chr.12: 13564036-13564036 on Build GRCh38
6209120 c.3005G>A p.G1002E Chr.12: 13564233-13564233 on Build GRCh38
401319 c.3026C>A p.P1009Q Chr.12: 13564212-13564212 on Build GRCh38
937462 c.3077G>A p.G1026D Chr.12: 13564161-13564161 on Build GRCh38
3980935 c.3115T>A p.Y1039N Chr.12: 13564123-13564123 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200355836 Chr.12: 13564205-13564205 on Build GRCh38
rs956362869 Chr.12: 13564210-13564210 on Build GRCh38
rs772837930 Chr.12: 13564238-13564238 on Build GRCh38
rs201866998 Chr.12: 13564092-13564092 on Build GRCh38
rs372040957 Chr.12: 13564142-13564142 on Build GRCh38
rs772364390 Chr.12: 13564064-13564064 on Build GRCh38
rs750257721 Chr.12: 13564118-13564118 on Build GRCh38
rs776986575 Chr.12: 13564079-13564079 on Build GRCh38
rs773762599 Chr.12: 13564233-13564233 on Build GRCh38
rs753715256 Chr.12: 13564124-13564124 on Build GRCh38
rs759983003 Chr.12: 13564164-13564164 on Build GRCh38
rs201439880 Chr.12: 13564226-13564226 on Build GRCh38
rs147762014 Chr.12: 13564121-13564121 on Build GRCh38
rs758734049 Chr.12: 13564064-13564065 on Build GRCh38
rs543452359 Chr.12: 13564082-13564082 on Build GRCh38
rs200265732 Chr.12: 13564182-13564182 on Build GRCh38
rs200210848 Chr.12: 13564202-13564202 on Build GRCh38
rs753613072 Chr.12: 13564249-13564249 on Build GRCh38
rs969024709 Chr.12: 13564240-13564240 on Build GRCh38
rs747431551 Chr.12: 13564187-13564187 on Build GRCh38
rs201029083 Chr.12: 13564101-13564101 on Build GRCh38
rs879254211 Chr.12: 13564043-13564043 on Build GRCh38
rs760531029 Chr.12: 13564241-13564241 on Build GRCh38
rs746372193 Chr.12: 13564225-13564225 on Build GRCh38
rs771609944 Chr.12: 13564073-13564073 on Build GRCh38
rs754878801 Chr.12: 13564127-13564127 on Build GRCh38
rs1048069070 Chr.12: 13564049-13564049 on Build GRCh38
rs1057518520 Chr.12: 13564228-13564229 on Build GRCh38
rs751021224 Chr.12: 13564100-13564100 on Build GRCh38
rs761281000 Chr.12: 13564234-13564234 on Build GRCh38
rs574130239 Chr.12: 13564195-13564195 on Build GRCh38
rs766092483 Chr.12: 13564247-13564247 on Build GRCh38
rs761084398 Chr.12: 13564069-13564069 on Build GRCh38
rs200887288 Chr.12: 13564157-13564157 on Build GRCh38
rs773737239 Chr.12: 13564068-13564068 on Build GRCh38
rs141109968 Chr.12: 13564191-13564191 on Build GRCh38
rs868013120 Chr.12: 13564103-13564103 on Build GRCh38
rs780435456 Chr.12: 13564067-13564068 on Build GRCh38
rs878854145 Chr.12: 13564148-13564150 on Build GRCh38
rs201963596 Chr.12: 13564162-13564162 on Build GRCh38
rs142071596 Chr.12: 13564058-13564058 on Build GRCh38
rs778873883 Chr.12: 13564133-13564133 on Build GRCh38
rs202222002 Chr.12: 13564120-13564120 on Build GRCh38
rs771413773 Chr.12: 13564188-13564188 on Build GRCh38
rs200621414 Chr.12: 13564061-13564061 on Build GRCh38
rs199769470 Chr.12: 13564232-13564232 on Build GRCh38
rs752072372 Chr.12: 13564070-13564071 on Build GRCh38
rs767114392 Chr.12: 13564112-13564112 on Build GRCh38
rs202109019 Chr.12: 13564200-13564200 on Build GRCh38
rs199857445 Chr.12: 13564095-13564095 on Build GRCh38
rs796052575 Chr.12: 13564139-13564139 on Build GRCh38
rs778114438 Chr.12: 13564170-13564170 on Build GRCh38

View Full Product Details