Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291916.1 | NP_001278845.1 | 1 - 1 | 81 | 71 |
NM_006938.3 | NP_008869.1 | 1 - 1 | 81 | 71 | |
GenBank mRNA | BC001721.1 | - | 1 - 1 | 42 | 71 |
BU565530.1 | - | 1 - 1 | 83 | 71 | |
HY043971.1 | - | 1 - 1 | 79 | 71 | |
J03798.1 | - | 1 - 1 | 70 | 71 |