Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 95844822-95845093 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTMR2
Entrez Gene ID
8898
Gene Name
myotubularin related protein 2
Gene Aliases
CMT4B, CMT4B1
UniGene
Hs.181326
Gene Chromosome Location
Chr.11: 95832880-95924207 on Build GRCh38
Transcripts
NM_001243571.1 NM_016156.5 NM_201278.2 NM_201281.2 XM_005274374.2 XM_005274375.2 XM_006718934.2 XM_006718935.2 XM_006718936.3 XM_011543058.2 XM_011543059.2 XM_017018517.1 XM_017018518.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4464116 c.1318C>T p.R440* Chr.11: 95845021-95845021 on Build GRCh38
933621 c.1052G>A p.R351H Chr.11: 95845071-95845071 on Build GRCh38
1258040 c.1363A>C p.S455R Chr.11: 95844976-95844976 on Build GRCh38
3810725 c.1083C>A p.L361L Chr.11: 95845040-95845040 on Build GRCh38
6716575 c.1159C>T p.R387* Chr.11: 95844964-95844964 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563602754 Chr.11: 95844913-95844913 on Build GRCh38
rs756723447 Chr.11: 95844939-95844939 on Build GRCh38
rs767737380 Chr.11: 95845036-95845036 on Build GRCh38
rs567520815 Chr.11: 95845026-95845026 on Build GRCh38
rs1023654455 Chr.11: 95844963-95844963 on Build GRCh38
rs141311831 Chr.11: 95845005-95845005 on Build GRCh38
rs758518766 Chr.11: 95844908-95844908 on Build GRCh38
rs563942287 Chr.11: 95844863-95844863 on Build GRCh38
rs371105248 Chr.11: 95844945-95844945 on Build GRCh38
rs1002342349 Chr.11: 95844880-95844880 on Build GRCh38
rs746686039 Chr.11: 95844959-95844959 on Build GRCh38
rs146572467 Chr.11: 95845003-95845003 on Build GRCh38
rs753138775 Chr.11: 95845040-95845040 on Build GRCh38
rs889511651 Chr.11: 95844852-95844852 on Build GRCh38
rs121434402 Chr.11: 95845063-95845063 on Build GRCh38
rs769693714 Chr.11: 95845000-95845000 on Build GRCh38
rs377452665 Chr.11: 95845062-95845062 on Build GRCh38
rs191940048 Chr.11: 95844904-95844904 on Build GRCh38
rs781632235 Chr.11: 95844926-95844926 on Build GRCh38
rs755287656 Chr.11: 95844923-95844923 on Build GRCh38
rs750523253 Chr.11: 95844907-95844907 on Build GRCh38
rs139949951 Chr.11: 95844911-95844911 on Build GRCh38
rs1041183729 Chr.11: 95844868-95844868 on Build GRCh38
rs766536590 Chr.11: 95845012-95845012 on Build GRCh38
rs139369561 Chr.11: 95844953-95844953 on Build GRCh38
rs372101052 Chr.11: 95844917-95844917 on Build GRCh38
rs374742138 Chr.11: 95845038-95845038 on Build GRCh38
rs751677491 Chr.11: 95845021-95845021 on Build GRCh38
rs776520645 Chr.11: 95844994-95844994 on Build GRCh38
rs114336434 Chr.11: 95844986-95844986 on Build GRCh38
rs377431314 Chr.11: 95844940-95844940 on Build GRCh38
rs371349038 Chr.11: 95845022-95845022 on Build GRCh38
rs199542109 Chr.11: 95845058-95845058 on Build GRCh38
rs768967010 Chr.11: 95844906-95844907 on Build GRCh38
rs748602901 Chr.11: 95844936-95844936 on Build GRCh38
rs565230483 Chr.11: 95844854-95844854 on Build GRCh38

View Full Product Details