Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000034.3 | NP_000025.1 | 3 - 4 | 309 | 102 |
GenBank mRNA | AK308495.1 | - | 3 - 4 | 228 | 102 |
BC004333.1 | - | 3 - 4 | 229 | 102 | |
DB057572.1 | - | 3 - 4 | 310 | 102 |