Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323281.1 | NP_001310210.1 | 9 - 10 | 1715 | 137 |
NM_001323282.1 | NP_001310211.1 | 10 - 11 | 1780 | 137 | |
NM_002895.4 | NP_002886.2 | 10 - 11 | 1642 | 137 | |
NM_183404.3 | NP_899662.1 | 10 - 11 | 1642 | 137 | |
XM_006723842.2 | XP_006723905.1 | 9 - 10 | 1353 | 137 | |
XM_011528955.2 | XP_011527257.1 | 10 - 11 | 1418 | 137 | |
XM_011528958.2 | XP_011527260.1 | 10 - 11 | 1418 | 137 | |
XM_017027992.1 | XP_016883481.1 | 11 - 12 | 1598 | 137 | |
GenBank mRNA | AK290380.1 | - | 10 - 11 | 1418 | 137 |
AK308096.1 | - | 10 - 11 | 1403 | 137 | |
BC032247.1 | - | 10 - 11 | 1425 | 137 | |
M74547.1 | - | 8 - 9 | 956 | 137 |