Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005469.3 | NP_005460.2 | 1 - 2 | 228 | 68 |
XM_011528480.2 | XP_011526782.1 | 1 - 2 | 190 | 68 | |
GenBank mRNA | AF014404.1 | - | 1 - 2 | 164 | 68 |
AF124264.1 | - | 1 - 2 | 134 | 68 | |
AK296973.1 | - | 1 - 2 | 216 | 68 | |
BC117155.1 | - | 1 - 2 | 156 | 68 | |
BC117157.1 | - | 1 - 2 | 156 | 68 | |
BI911038.1 | - | 1 - 2 | 214 | 68 | |
CR541771.1 | - | 1 - 2 | 134 | 68 | |
CR541778.1 | - | 1 - 2 | 134 | 68 | |
DB470803.3 | - | 1 - 2 | 228 | 68 | |
X86032.1 | - | 1 - 2 | 202 | 68 |