Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42752320-42752583 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTBK2
Entrez Gene ID
146057
Gene Name
tau tubulin kinase 2
Gene Aliases
SCA11, TTBK
UniGene
Hs.646511
Gene Chromosome Location
Chr.15: 42738974-42920995 on Build GRCh38
Transcripts
NM_173500.3 XM_005254171.4 XM_005254173.4 XM_006720402.3 XM_006720403.3 XM_017021950.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
700552 c.2683G>C p.D895H Chr.15: 42752563-42752563 on Build GRCh38
5909475 c.2681G>A p.G894E Chr.15: 42752565-42752565 on Build GRCh38
5952240 c.2789G>A p.R930Q Chr.15: 42752457-42752457 on Build GRCh38
4539667 c.2719G>A p.E907K Chr.15: 42752527-42752527 on Build GRCh38
5628421 c.2671G>C p.G891R Chr.15: 42752575-42752575 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs532401842 Chr.15: 42752424-42752424 on Build GRCh38
rs933276422 Chr.15: 42752561-42752561 on Build GRCh38
rs771977308 Chr.15: 42752439-42752439 on Build GRCh38
rs765811535 Chr.15: 42752367-42752367 on Build GRCh38
rs535040519 Chr.15: 42752560-42752560 on Build GRCh38
rs574547214 Chr.15: 42752396-42752396 on Build GRCh38
rs374324347 Chr.15: 42752428-42752428 on Build GRCh38
rs757518859 Chr.15: 42752363-42752363 on Build GRCh38
rs944447124 Chr.15: 42752440-42752440 on Build GRCh38
rs755069708 Chr.15: 42752533-42752533 on Build GRCh38
rs371858440 Chr.15: 42752459-42752459 on Build GRCh38
rs552522869 Chr.15: 42752489-42752489 on Build GRCh38
rs201348630 Chr.15: 42752415-42752415 on Build GRCh38
rs760324636 Chr.15: 42752346-42752346 on Build GRCh38
rs781516018 Chr.15: 42752420-42752420 on Build GRCh38
rs1022933283 Chr.15: 42752364-42752364 on Build GRCh38
rs368843591 Chr.15: 42752398-42752398 on Build GRCh38
rs745618805 Chr.15: 42752559-42752559 on Build GRCh38
rs371807025 Chr.15: 42752500-42752500 on Build GRCh38
rs974820996 Chr.15: 42752518-42752518 on Build GRCh38
rs1040549147 Chr.15: 42752443-42752443 on Build GRCh38
rs987302563 Chr.15: 42752490-42752490 on Build GRCh38
rs774994835 Chr.15: 42752442-42752442 on Build GRCh38
rs767042342 Chr.15: 42752457-42752457 on Build GRCh38
rs765923162 Chr.15: 42752349-42752349 on Build GRCh38
rs368034421 Chr.15: 42752480-42752480 on Build GRCh38
rs866877821 Chr.15: 42752350-42752350 on Build GRCh38
rs368262141 Chr.15: 42752448-42752448 on Build GRCh38
rs751008854 Chr.15: 42752485-42752485 on Build GRCh38
rs376245132 Chr.15: 42752479-42752479 on Build GRCh38
rs776231019 Chr.15: 42752470-42752470 on Build GRCh38
rs751712578 Chr.15: 42752517-42752517 on Build GRCh38
rs754220583 Chr.15: 42752357-42752357 on Build GRCh38
rs374795359 Chr.15: 42752558-42752558 on Build GRCh38
rs540476257 Chr.15: 42752399-42752399 on Build GRCh38
rs751131823 Chr.15: 42752386-42752386 on Build GRCh38
rs972275430 Chr.15: 42752355-42752355 on Build GRCh38
rs964464006 Chr.15: 42752516-42752516 on Build GRCh38
rs781607577 Chr.15: 42752534-42752534 on Build GRCh38
rs745787994 Chr.15: 42752432-42752432 on Build GRCh38
rs750869234 Chr.15: 42752370-42752370 on Build GRCh38
rs886042912 Chr.15: 42752387-42752387 on Build GRCh38
rs757141457 Chr.15: 42752556-42752556 on Build GRCh38
rs758567031 Chr.15: 42752382-42752382 on Build GRCh38
rs760128494 Chr.15: 42752451-42752451 on Build GRCh38
rs752842969 Chr.15: 42752535-42752535 on Build GRCh38

View Full Product Details