Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 166199128-166199381 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101929680
Entrez Gene ID
101929680
Gene Name
uncharacterized LOC101929680
Gene Aliases
-
UniGene
Hs.662210
Gene Chromosome Location
Chr.2: 166081531-166301784 on Build GRCh38
Transcripts
NR_110260.1 NM_002977.3 XM_005246757.2 XM_011511616.2 XM_011511617.2 XM_011511618.2 XM_017004668.1 XM_017004669.1

Gene Symbol
SCN9A
Entrez Gene ID
6335
Gene Name
sodium voltage-gated channel alpha subunit 9
Gene Aliases
ETHA, FEB3B, GEFSP7, HSAN2D, NE-NA, NENA, Nav1.7, PN1, SFNP
UniGene
Hs.439145
Gene Chromosome Location
Chr.2: 166195185-166375987 on Build GRCh38
Transcripts
NR_110260.1 NM_002977.3 XM_005246757.2 XM_011511616.2 XM_011511617.2 XM_011511618.2 XM_017004668.1 XM_017004669.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1008763 c.5317G>A p.E1773K Chr.2: 166199289-166199289 on Build GRCh38
6509958 c.5341G>T p.D1781Y Chr.2: 166199265-166199265 on Build GRCh38
5394777 c.5467C>T p.Q1823* Chr.2: 166199175-166199175 on Build GRCh38
3406998 c.5482G>T p.D1828Y Chr.2: 166199160-166199160 on Build GRCh38
4763399 c.5468A>C p.Q1823P Chr.2: 166199174-166199174 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200050326 Chr.2: 166199288-166199288 on Build GRCh38
rs751881997 Chr.2: 166199336-166199336 on Build GRCh38
rs200745478 Chr.2: 166199246-166199246 on Build GRCh38
rs762575742 Chr.2: 166199151-166199151 on Build GRCh38
rs1021711301 Chr.2: 166199181-166199181 on Build GRCh38
rs751325279 Chr.2: 166199152-166199152 on Build GRCh38
rs199954555 Chr.2: 166199214-166199214 on Build GRCh38
rs200803311 Chr.2: 166199149-166199149 on Build GRCh38
rs1017717952 Chr.2: 166199278-166199278 on Build GRCh38
rs895083256 Chr.2: 166199341-166199341 on Build GRCh38
rs1012108087 Chr.2: 166199179-166199179 on Build GRCh38
rs879254324 Chr.2: 166199219-166199219 on Build GRCh38
rs756016813 Chr.2: 166199180-166199180 on Build GRCh38
rs1013397100 Chr.2: 166199356-166199356 on Build GRCh38
rs750503411 Chr.2: 166199196-166199196 on Build GRCh38
rs764035131 Chr.2: 166199313-166199313 on Build GRCh38
rs747367937 Chr.2: 166199227-166199227 on Build GRCh38
rs778928947 Chr.2: 166199183-166199183 on Build GRCh38
rs199696636 Chr.2: 166199290-166199290 on Build GRCh38
rs760665758 Chr.2: 166199307-166199307 on Build GRCh38
rs762462693 Chr.2: 166199265-166199265 on Build GRCh38
rs973563108 Chr.2: 166199317-166199317 on Build GRCh38
rs781023511 Chr.2: 166199314-166199314 on Build GRCh38
rs752580340 Chr.2: 166199172-166199172 on Build GRCh38
rs770494883 Chr.2: 166199284-166199284 on Build GRCh38
rs759259640 Chr.2: 166199165-166199165 on Build GRCh38
rs770416478 Chr.2: 166199247-166199247 on Build GRCh38
rs780166088 Chr.2: 166199329-166199329 on Build GRCh38
rs759347599 Chr.2: 166199293-166199293 on Build GRCh38
rs748702835 Chr.2: 166199236-166199236 on Build GRCh38
rs769097206 Chr.2: 166199229-166199229 on Build GRCh38
rs767347325 Chr.2: 166199295-166199295 on Build GRCh38
rs936563850 Chr.2: 166199281-166199281 on Build GRCh38
rs113490809 Chr.2: 166199252-166199252 on Build GRCh38
rs945489250 Chr.2: 166199230-166199231 on Build GRCh38
rs201196474 Chr.2: 166199305-166199305 on Build GRCh38
rs986047440 Chr.2: 166199266-166199266 on Build GRCh38
rs758469073 Chr.2: 166199326-166199326 on Build GRCh38
rs201972510 Chr.2: 166199166-166199166 on Build GRCh38
rs201875421 Chr.2: 166199260-166199260 on Build GRCh38
rs200196731 Chr.2: 166199148-166199148 on Build GRCh38
rs755222805 Chr.2: 166199354-166199354 on Build GRCh38
rs372311962 Chr.2: 166199231-166199231 on Build GRCh38
rs768621572 Chr.2: 166199253-166199253 on Build GRCh38

View Full Product Details