Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100404872-100405143 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZCWPW1
Entrez Gene ID
55063
Gene Name
zinc finger CW-type and PWWP domain containing 1
Gene Aliases
ZCW1
UniGene
Hs.105191
Gene Chromosome Location
Chr.7: 100400826-100429239 on Build GRCh38
Transcripts
NM_001258008.1 NM_017984.4 XM_005250479.2 XM_005250480.2 XM_006716035.3 XM_006716036.2 XM_006716037.3 XM_006716038.3 XM_006716040.3 XM_017012379.1 XM_017012380.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6530193 c.1249C>G p.Q417E Chr.7: 100405015-100405015 on Build GRCh38
7146462 c.1246A>G p.I416V Chr.7: 100405018-100405018 on Build GRCh38
7416682 c.1218G>A p.M406I Chr.7: 100405046-100405046 on Build GRCh38
6826016 c.1219A>G p.M407V Chr.7: 100405045-100405045 on Build GRCh38
5683833 c.1206G>T p.G402G Chr.7: 100405058-100405058 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs538555436 Chr.7: 100405059-100405059 on Build GRCh38
rs753016718 Chr.7: 100404995-100404995 on Build GRCh38
rs768000707 Chr.7: 100404990-100404990 on Build GRCh38
rs534404386 Chr.7: 100405025-100405025 on Build GRCh38
rs372293650 Chr.7: 100405014-100405014 on Build GRCh38
rs973711267 Chr.7: 100405106-100405106 on Build GRCh38
rs890855803 Chr.7: 100404965-100404965 on Build GRCh38
rs751636333 Chr.7: 100404980-100404980 on Build GRCh38
rs763053397 Chr.7: 100404962-100404962 on Build GRCh38
rs571490957 Chr.7: 100404903-100404903 on Build GRCh38
rs555639782 Chr.7: 100405086-100405086 on Build GRCh38
rs756945353 Chr.7: 100405038-100405038 on Build GRCh38
rs753207257 Chr.7: 100405100-100405100 on Build GRCh38
rs755496338 Chr.7: 100404983-100404983 on Build GRCh38
rs182080895 Chr.7: 100404963-100404963 on Build GRCh38
rs999736535 Chr.7: 100404977-100404977 on Build GRCh38
rs779988689 Chr.7: 100405064-100405064 on Build GRCh38
rs79146967 Chr.7: 100405087-100405087 on Build GRCh38
rs763213220 Chr.7: 100405084-100405084 on Build GRCh38
rs759602275 Chr.7: 100405094-100405094 on Build GRCh38
rs770873183 Chr.7: 100404958-100404959 on Build GRCh38
rs903775560 Chr.7: 100404934-100404934 on Build GRCh38
rs768402660 Chr.7: 100405080-100405080 on Build GRCh38
rs761229659 Chr.7: 100405105-100405105 on Build GRCh38
rs757267690 Chr.7: 100405037-100405038 on Build GRCh38
rs776380737 Chr.7: 100405083-100405083 on Build GRCh38
rs778340456 Chr.7: 100405042-100405042 on Build GRCh38
rs745376116 Chr.7: 100405050-100405050 on Build GRCh38
rs777598413 Chr.7: 100405023-100405023 on Build GRCh38
rs1022183057 Chr.7: 100404925-100404925 on Build GRCh38
rs746805431 Chr.7: 100405070-100405070 on Build GRCh38
rs1050763420 Chr.7: 100404939-100404939 on Build GRCh38
rs767929906 Chr.7: 100405095-100405095 on Build GRCh38
rs771911580 Chr.7: 100405057-100405057 on Build GRCh38

View Full Product Details