Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100627570-100627795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TFR2
Entrez Gene ID
7036
Gene Name
transferrin receptor 2
Gene Aliases
HFE3, TFRC2
UniGene
Hs.544932
Gene Chromosome Location
Chr.7: 100620416-100642780 on Build GRCh38
Transcripts
NM_001206855.1 NM_003227.3 XM_005250553.4 XM_017012573.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3080551 c.1750G>A p.E584K Chr.7: 100627594-100627594 on Build GRCh38
6800604 c.1761T>A p.F587L Chr.7: 100627583-100627583 on Build GRCh38
1446602 c.1719C>A p.S573S Chr.7: 100627625-100627625 on Build GRCh38
6027438 c.1713C>A p.A571A Chr.7: 100627631-100627631 on Build GRCh38
6800602 c.1746C>T p.A582A Chr.7: 100627598-100627598 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200217701 Chr.7: 100627674-100627674 on Build GRCh38
rs749077194 Chr.7: 100627706-100627706 on Build GRCh38
rs769477274 Chr.7: 100627604-100627604 on Build GRCh38
rs766102069 Chr.7: 100627678-100627678 on Build GRCh38
rs775586229 Chr.7: 100627643-100627643 on Build GRCh38
rs746098454 Chr.7: 100627632-100627632 on Build GRCh38
rs566516530 Chr.7: 100627731-100627731 on Build GRCh38
rs780778077 Chr.7: 100627684-100627684 on Build GRCh38
rs760739518 Chr.7: 100627654-100627654 on Build GRCh38
rs370375593 Chr.7: 100627597-100627597 on Build GRCh38
rs758038845 Chr.7: 100627676-100627676 on Build GRCh38
rs531726662 Chr.7: 100627657-100627657 on Build GRCh38
rs80338887 Chr.7: 100627761-100627761 on Build GRCh38
rs963878047 Chr.7: 100627680-100627680 on Build GRCh38
rs973805993 Chr.7: 100627702-100627702 on Build GRCh38
rs776654518 Chr.7: 100627726-100627726 on Build GRCh38
rs563694182 Chr.7: 100627730-100627730 on Build GRCh38
rs953857568 Chr.7: 100627700-100627700 on Build GRCh38
rs751623769 Chr.7: 100627748-100627748 on Build GRCh38
rs772314714 Chr.7: 100627642-100627642 on Build GRCh38
rs751605127 Chr.7: 100627729-100627730 on Build GRCh38
rs781258602 Chr.7: 100627598-100627598 on Build GRCh38
rs748036801 Chr.7: 100627602-100627602 on Build GRCh38
rs754545158 Chr.7: 100627683-100627683 on Build GRCh38
rs751258355 Chr.7: 100627734-100627734 on Build GRCh38
rs539310085 Chr.7: 100627709-100627709 on Build GRCh38
rs764366227 Chr.7: 100627653-100627654 on Build GRCh38
rs756044574 Chr.7: 100627740-100627740 on Build GRCh38
rs764643944 Chr.7: 100627656-100627656 on Build GRCh38
rs35383296 Chr.7: 100627682-100627682 on Build GRCh38
rs144723097 Chr.7: 100627619-100627619 on Build GRCh38
rs762251269 Chr.7: 100627730-100627730 on Build GRCh38
rs765554083 Chr.7: 100627668-100627668 on Build GRCh38
rs41295909 Chr.7: 100627704-100627704 on Build GRCh38
rs916073801 Chr.7: 100627772-100627772 on Build GRCh38
rs112165576 Chr.7: 100627663-100627663 on Build GRCh38
rs560458124 Chr.7: 100627646-100627646 on Build GRCh38
rs752709708 Chr.7: 100627685-100627685 on Build GRCh38
rs756027634 Chr.7: 100627696-100627696 on Build GRCh38
rs200813529 Chr.7: 100627723-100627723 on Build GRCh38
rs779989093 Chr.7: 100627771-100627773 on Build GRCh38
rs201936620 Chr.7: 100627665-100627665 on Build GRCh38
rs41295906 Chr.7: 100627735-100627735 on Build GRCh38

View Full Product Details