Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100406609-100406882 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZCWPW1
Entrez Gene ID
55063
Gene Name
zinc finger CW-type and PWWP domain containing 1
Gene Aliases
ZCW1
UniGene
Hs.105191
Gene Chromosome Location
Chr.7: 100400826-100429239 on Build GRCh38
Transcripts
NM_001258008.1 NM_017984.4 XM_005250479.2 XM_005250480.2 XM_006716035.3 XM_006716036.2 XM_006716037.3 XM_006716038.3 XM_006716040.3 XM_017012379.1 XM_017012380.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3630737 c.1116C>T p.I372I Chr.7: 100406748-100406748 on Build GRCh38
5037010 c.1170+3G>A p.? Chr.7: 100406691-100406691 on Build GRCh38
3630736 c.1118C>A p.P373Q Chr.7: 100406746-100406746 on Build GRCh38
1082838 c.1103C>A p.S368Y Chr.7: 100406761-100406761 on Build GRCh38
4918062 c.1158G>A p.E386E Chr.7: 100406706-100406706 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757536331 Chr.7: 100406781-100406781 on Build GRCh38
rs781042890 Chr.7: 100406737-100406737 on Build GRCh38
rs201574533 Chr.7: 100406758-100406758 on Build GRCh38
rs777232712 Chr.7: 100406801-100406801 on Build GRCh38
rs781022769 Chr.7: 100406658-100406658 on Build GRCh38
rs201240514 Chr.7: 100406665-100406665 on Build GRCh38
rs986072859 Chr.7: 100406836-100406836 on Build GRCh38
rs754976331 Chr.7: 100406735-100406735 on Build GRCh38
rs769303827 Chr.7: 100406797-100406797 on Build GRCh38
rs1476679 Chr.7: 100406823-100406823 on Build GRCh38
rs768560435 Chr.7: 100406655-100406655 on Build GRCh38
rs764824841 Chr.7: 100406713-100406713 on Build GRCh38
rs762214750 Chr.7: 100406711-100406711 on Build GRCh38
rs369290615 Chr.7: 100406716-100406716 on Build GRCh38
rs911215576 Chr.7: 100406861-100406861 on Build GRCh38
rs527408897 Chr.7: 100406733-100406733 on Build GRCh38
rs373166304 Chr.7: 100406717-100406717 on Build GRCh38
rs755915575 Chr.7: 100406722-100406727 on Build GRCh38
rs779665911 Chr.7: 100406729-100406729 on Build GRCh38
rs754255192 Chr.7: 100406709-100406709 on Build GRCh38
rs773428981 Chr.7: 100406844-100406844 on Build GRCh38
rs147445817 Chr.7: 100406703-100406703 on Build GRCh38
rs759563877 Chr.7: 100406693-100406693 on Build GRCh38
rs746990435 Chr.7: 100406645-100406645 on Build GRCh38
rs758061043 Chr.7: 100406755-100406755 on Build GRCh38
rs6970350 Chr.7: 100406771-100406771 on Build GRCh38
rs771154657 Chr.7: 100406678-100406678 on Build GRCh38
rs747958089 Chr.7: 100406754-100406754 on Build GRCh38
rs761319706 Chr.7: 100406700-100406700 on Build GRCh38
rs781741302 Chr.7: 100406782-100406782 on Build GRCh38
rs774584375 Chr.7: 100406685-100406685 on Build GRCh38
rs762302590 Chr.7: 100406813-100406813 on Build GRCh38
rs745646119 Chr.7: 100406661-100406663 on Build GRCh38
rs200331453 Chr.7: 100406696-100406696 on Build GRCh38
rs756485668 Chr.7: 100406766-100406766 on Build GRCh38

View Full Product Details