Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27469764-27469999 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA2
Entrez Gene ID
1135
Gene Name
cholinergic receptor nicotinic alpha 2 subunit
Gene Aliases
-
UniGene
Hs.57718
Gene Chromosome Location
Chr.8: 27459761-27479296 on Build GRCh38
Transcripts
NM_000742.3 NM_001282455.1 XM_005273397.2 XM_006716282.1 XM_011544388.1 XM_011544389.1 XM_017013003.1 XM_017013004.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4996940 c.242C>T p.S81L Chr.8: 27469813-27469813 on Build GRCh38
4448654 c.113delG p.G38fs*47 Chr.8: 27469942-27469942 on Build GRCh38
4959822 c.278C>T p.A93V Chr.8: 27469777-27469777 on Build GRCh38
4366884 c.109C>T p.P37S Chr.8: 27469946-27469946 on Build GRCh38
1098743 c.92G>A p.R31H Chr.8: 27469963-27469963 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749670139 Chr.8: 27469798-27469798 on Build GRCh38
rs763260906 Chr.8: 27469974-27469974 on Build GRCh38
rs548268816 Chr.8: 27469852-27469852 on Build GRCh38
rs889869424 Chr.8: 27469893-27469893 on Build GRCh38
rs778838030 Chr.8: 27469924-27469924 on Build GRCh38
rs1016586819 Chr.8: 27469921-27469921 on Build GRCh38
rs745670660 Chr.8: 27469936-27469936 on Build GRCh38
rs776191995 Chr.8: 27469963-27469963 on Build GRCh38
rs148768097 Chr.8: 27469872-27469872 on Build GRCh38
rs748345482 Chr.8: 27469918-27469918 on Build GRCh38
rs151268950 Chr.8: 27469882-27469882 on Build GRCh38
rs765581867 Chr.8: 27469896-27469896 on Build GRCh38
rs766863642 Chr.8: 27469977-27469977 on Build GRCh38
rs770949841 Chr.8: 27469832-27469832 on Build GRCh38
rs773501890 Chr.8: 27469965-27469965 on Build GRCh38
rs758848783 Chr.8: 27469905-27469905 on Build GRCh38
rs200035862 Chr.8: 27469862-27469862 on Build GRCh38
rs568479156 Chr.8: 27469873-27469873 on Build GRCh38
rs868195779 Chr.8: 27469888-27469888 on Build GRCh38
rs139552324 Chr.8: 27469886-27469886 on Build GRCh38
rs757700781 Chr.8: 27469808-27469808 on Build GRCh38
rs772119766 Chr.8: 27469823-27469823 on Build GRCh38
rs532800808 Chr.8: 27469819-27469819 on Build GRCh38
rs144185168 Chr.8: 27469889-27469889 on Build GRCh38
rs749040383 Chr.8: 27469866-27469866 on Build GRCh38
rs771956737 Chr.8: 27469880-27469880 on Build GRCh38
rs770203806 Chr.8: 27469919-27469919 on Build GRCh38
rs369806823 Chr.8: 27469833-27469833 on Build GRCh38
rs376970816 Chr.8: 27469853-27469853 on Build GRCh38
rs759538013 Chr.8: 27469877-27469877 on Build GRCh38
rs747073463 Chr.8: 27469858-27469858 on Build GRCh38
rs767104640 Chr.8: 27469839-27469839 on Build GRCh38
rs752371632 Chr.8: 27469785-27469785 on Build GRCh38
rs903870658 Chr.8: 27469871-27469871 on Build GRCh38
rs759595350 Chr.8: 27469834-27469834 on Build GRCh38
rs796052310 Chr.8: 27469934-27469934 on Build GRCh38
rs201890355 Chr.8: 27469841-27469841 on Build GRCh38
rs762847144 Chr.8: 27469831-27469831 on Build GRCh38
rs538453326 Chr.8: 27469955-27469955 on Build GRCh38
rs201922955 Chr.8: 27469840-27469840 on Build GRCh38
rs755757952 Chr.8: 27469786-27469786 on Build GRCh38
rs199810678 Chr.8: 27469828-27469828 on Build GRCh38
rs146763552 Chr.8: 27469906-27469906 on Build GRCh38
rs142375828 Chr.8: 27469874-27469874 on Build GRCh38
rs771977388 Chr.8: 27469949-27469949 on Build GRCh38
rs371737919 Chr.8: 27469830-27469830 on Build GRCh38
rs867006598 Chr.8: 27469931-27469931 on Build GRCh38
rs778191765 Chr.8: 27469796-27469796 on Build GRCh38
rs747017890 Chr.8: 27469960-27469960 on Build GRCh38
rs796052307 Chr.8: 27469934-27469934 on Build GRCh38
rs764887214 Chr.8: 27469964-27469964 on Build GRCh38
rs79290097 Chr.8: 27469843-27469843 on Build GRCh38
rs532212433 Chr.8: 27469795-27469795 on Build GRCh38
rs775319340 Chr.8: 27469957-27469957 on Build GRCh38
rs200277900 Chr.8: 27469827-27469827 on Build GRCh38
rs74772771 Chr.8: 27469915-27469915 on Build GRCh38
rs761493925 Chr.8: 27469894-27469894 on Build GRCh38
rs768021745 Chr.8: 27469962-27469962 on Build GRCh38
rs553897766 Chr.8: 27469897-27469897 on Build GRCh38
rs200729883 Chr.8: 27469887-27469887 on Build GRCh38
rs144316375 Chr.8: 27469845-27469845 on Build GRCh38
rs201244136 Chr.8: 27469809-27469809 on Build GRCh38
rs559859378 Chr.8: 27469864-27469864 on Build GRCh38

View Full Product Details