Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 23837298-23837523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRKCB
Entrez Gene ID
5579
Gene Name
protein kinase C beta
Gene Aliases
PKC-beta, PKCB, PRKCB1, PRKCB2
UniGene
Hs.460355
Gene Chromosome Location
Chr.16: 23835979-24220611 on Build GRCh38
Transcripts
NM_002738.6 NM_212535.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3988356 c.177C>T p.G59G Chr.16: 23837378-23837378 on Build GRCh38
1323643 c.199T>A p.C67S Chr.16: 23837400-23837400 on Build GRCh38
1323644 c.199T>A p.C67S Chr.16: 23837400-23837400 on Build GRCh38
6428816 c.179T>C p.F60S Chr.16: 23837380-23837380 on Build GRCh38
7108239 c.174-1G>T p.? Chr.16: 23837374-23837374 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746031424 Chr.16: 23837414-23837415 on Build GRCh38
rs3729884 Chr.16: 23837491-23837491 on Build GRCh38
rs769737298 Chr.16: 23837387-23837387 on Build GRCh38
rs766294893 Chr.16: 23837416-23837416 on Build GRCh38
rs746668304 Chr.16: 23837341-23837341 on Build GRCh38
rs776139953 Chr.16: 23837342-23837342 on Build GRCh38
rs912429510 Chr.16: 23837426-23837426 on Build GRCh38
rs538173627 Chr.16: 23837465-23837465 on Build GRCh38
rs779340232 Chr.16: 23837327-23837327 on Build GRCh38
rs764223724 Chr.16: 23837434-23837434 on Build GRCh38
rs757511512 Chr.16: 23837329-23837330 on Build GRCh38
rs868688793 Chr.16: 23837402-23837402 on Build GRCh38
rs551641254 Chr.16: 23837498-23837498 on Build GRCh38
rs866469671 Chr.16: 23837391-23837391 on Build GRCh38
rs752903660 Chr.16: 23837424-23837424 on Build GRCh38
rs758721130 Chr.16: 23837449-23837449 on Build GRCh38
rs1040370807 Chr.16: 23837417-23837417 on Build GRCh38
rs746212043 Chr.16: 23837333-23837333 on Build GRCh38
rs569130040 Chr.16: 23837444-23837444 on Build GRCh38
rs778841260 Chr.16: 23837352-23837353 on Build GRCh38
rs761275938 Chr.16: 23837360-23837360 on Build GRCh38
rs3729883 Chr.16: 23837441-23837441 on Build GRCh38
rs761013047 Chr.16: 23837433-23837433 on Build GRCh38
rs758875219 Chr.16: 23837439-23837439 on Build GRCh38
rs750951281 Chr.16: 23837446-23837446 on Build GRCh38
rs779259583 Chr.16: 23837468-23837468 on Build GRCh38
rs779623485 Chr.16: 23837339-23837339 on Build GRCh38
rs780530354 Chr.16: 23837414-23837414 on Build GRCh38
rs768966596 Chr.16: 23837463-23837463 on Build GRCh38

View Full Product Details