Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23417160-23417385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MHRT
Entrez Gene ID
104564225
Gene Name
myosin heavy chain-associated RNA transcript
Gene Aliases
Myheart
UniGene
Gene Chromosome Location
Chr.14: 23415450-23417595 on Build GRCh38
Transcripts
NR_126491.1 NM_000257.3 XM_017021340.1

Gene Symbol
MYH7
Entrez Gene ID
4625
Gene Name
myosin, heavy chain 7, cardiac muscle, beta
Gene Aliases
CMD1S, CMH1, MPD1, MYHCB, SPMD, SPMM
UniGene
Hs.719946
Gene Chromosome Location
Chr.14: 23412738-23435686 on Build GRCh38
Transcripts
NR_126491.1 NM_000257.3 XM_017021340.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3987586 c.4459G>A p.A1487T Chr.14: 23417213-23417213 on Build GRCh38
6521353 c.4423C>T p.R1475C Chr.14: 23417249-23417249 on Build GRCh38
5790673 c.4501G>A p.E1501K Chr.14: 23417171-23417171 on Build GRCh38
416234 c.4486G>A p.E1496K Chr.14: 23417186-23417186 on Build GRCh38
6521352 c.4500G>T p.R1500R Chr.14: 23417172-23417172 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs397516213 Chr.14: 23417277-23417277 on Build GRCh38
rs763343366 Chr.14: 23417201-23417201 on Build GRCh38
rs727505281 Chr.14: 23417310-23417310 on Build GRCh38
rs2754156 Chr.14: 23417352-23417352 on Build GRCh38
rs200855376 Chr.14: 23417216-23417216 on Build GRCh38
rs766354665 Chr.14: 23417350-23417350 on Build GRCh38
rs748012379 Chr.14: 23417365-23417365 on Build GRCh38
rs397516214 Chr.14: 23417273-23417273 on Build GRCh38
rs950409210 Chr.14: 23417323-23417323 on Build GRCh38
rs992526361 Chr.14: 23417196-23417196 on Build GRCh38
rs587779414 Chr.14: 23417230-23417230 on Build GRCh38
rs772562745 Chr.14: 23417299-23417299 on Build GRCh38
rs753814030 Chr.14: 23417347-23417347 on Build GRCh38
rs876657884 Chr.14: 23417270-23417270 on Build GRCh38
rs767666398 Chr.14: 23417231-23417231 on Build GRCh38
rs764410124 Chr.14: 23417339-23417339 on Build GRCh38
rs397516215 Chr.14: 23417264-23417264 on Build GRCh38
rs1033511138 Chr.14: 23417278-23417278 on Build GRCh38
rs757428727 Chr.14: 23417349-23417349 on Build GRCh38
rs727505329 Chr.14: 23417191-23417191 on Build GRCh38
rs781203524 Chr.14: 23417283-23417283 on Build GRCh38
rs948329167 Chr.14: 23417209-23417209 on Build GRCh38
rs755412978 Chr.14: 23417363-23417363 on Build GRCh38
rs890401818 Chr.14: 23417284-23417284 on Build GRCh38
rs543141581 Chr.14: 23417328-23417328 on Build GRCh38
rs771377530 Chr.14: 23417308-23417308 on Build GRCh38
rs397516217 Chr.14: 23417236-23417236 on Build GRCh38
rs578166720 Chr.14: 23417262-23417262 on Build GRCh38
rs995126992 Chr.14: 23417290-23417290 on Build GRCh38
rs928227757 Chr.14: 23417183-23417183 on Build GRCh38
rs754829218 Chr.14: 23417282-23417282 on Build GRCh38
rs876661373 Chr.14: 23417309-23417309 on Build GRCh38
rs558413189 Chr.14: 23417223-23417223 on Build GRCh38
rs190577943 Chr.14: 23417300-23417300 on Build GRCh38
rs368078397 Chr.14: 23417324-23417324 on Build GRCh38
rs764058571 Chr.14: 23417256-23417256 on Build GRCh38
rs529657543 Chr.14: 23417252-23417252 on Build GRCh38
rs766909770 Chr.14: 23417213-23417213 on Build GRCh38
rs3729823 Chr.14: 23417200-23417200 on Build GRCh38
rs752867429 Chr.14: 23417237-23417237 on Build GRCh38
rs2854801 Chr.14: 23417330-23417330 on Build GRCh38
rs61737803 Chr.14: 23417220-23417220 on Build GRCh38
rs397516218 Chr.14: 23417185-23417185 on Build GRCh38
rs571704020 Chr.14: 23417214-23417214 on Build GRCh38
rs727504734 Chr.14: 23417320-23417320 on Build GRCh38
rs730880911 Chr.14: 23417202-23417202 on Build GRCh38
rs781107230 Chr.14: 23417364-23417364 on Build GRCh38
rs397516216 Chr.14: 23417261-23417261 on Build GRCh38
rs139646545 Chr.14: 23417249-23417249 on Build GRCh38
rs201307101 Chr.14: 23417295-23417295 on Build GRCh38
rs370093487 Chr.14: 23417325-23417325 on Build GRCh38

View Full Product Details