Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 95847561-95847798 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTMR2
Entrez Gene ID
8898
Gene Name
myotubularin related protein 2
Gene Aliases
CMT4B, CMT4B1
UniGene
Hs.181326
Gene Chromosome Location
Chr.11: 95832880-95924207 on Build GRCh38
Transcripts
NM_001243571.1 NM_016156.5 NM_201278.2 NM_201281.2 XM_005274374.2 XM_005274375.2 XM_006718934.2 XM_006718935.2 XM_006718936.3 XM_011543058.2 XM_011543059.2 XM_017018517.1 XM_017018518.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
236163 c.909G>T p.E303D Chr.11: 95847768-95847768 on Build GRCh38
236164 c.1125G>T p.E375D Chr.11: 95847768-95847768 on Build GRCh38
4404045 c.1158T>G p.T386T Chr.11: 95847735-95847735 on Build GRCh38
3398227 c.930C>T p.N310N Chr.11: 95847747-95847747 on Build GRCh38
3398226 c.1146C>T p.N382N Chr.11: 95847747-95847747 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879248604 Chr.11: 95847771-95847771 on Build GRCh38
rs770926882 Chr.11: 95847667-95847667 on Build GRCh38
rs1033228220 Chr.11: 95847647-95847647 on Build GRCh38
rs754494874 Chr.11: 95847711-95847711 on Build GRCh38
rs760992672 Chr.11: 95847688-95847688 on Build GRCh38
rs764511665 Chr.11: 95847697-95847697 on Build GRCh38
rs774208121 Chr.11: 95847671-95847671 on Build GRCh38
rs923304861 Chr.11: 95847635-95847635 on Build GRCh38
rs775320002 Chr.11: 95847684-95847687 on Build GRCh38
rs765744071 Chr.11: 95847704-95847704 on Build GRCh38
rs777156783 Chr.11: 95847698-95847698 on Build GRCh38
rs369314971 Chr.11: 95847680-95847680 on Build GRCh38
rs527341607 Chr.11: 95847714-95847714 on Build GRCh38
rs114883605 Chr.11: 95847772-95847772 on Build GRCh38
rs752277785 Chr.11: 95847725-95847725 on Build GRCh38
rs775843199 Chr.11: 95847685-95847685 on Build GRCh38
rs919359093 Chr.11: 95847710-95847710 on Build GRCh38
rs762132622 Chr.11: 95847700-95847700 on Build GRCh38
rs566204 Chr.11: 95847762-95847762 on Build GRCh38
rs749090978 Chr.11: 95847764-95847764 on Build GRCh38
rs1043569332 Chr.11: 95847768-95847768 on Build GRCh38
rs541804986 Chr.11: 95847610-95847610 on Build GRCh38
rs988897482 Chr.11: 95847743-95847743 on Build GRCh38
rs755797793 Chr.11: 95847750-95847750 on Build GRCh38
rs774043555 Chr.11: 95847643-95847644 on Build GRCh38
rs375029987 Chr.11: 95847665-95847665 on Build GRCh38
rs745929770 Chr.11: 95847675-95847675 on Build GRCh38
rs750955214 Chr.11: 95847707-95847707 on Build GRCh38
rs199736240 Chr.11: 95847723-95847723 on Build GRCh38

View Full Product Details