Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50582110-50582351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHKB
Entrez Gene ID
1120
Gene Name
choline kinase beta
Gene Aliases
CHETK, CHKL, CK, CKB, CKEKB, EK, EKB, MDCMC
UniGene
Hs.654827
Gene Chromosome Location
Chr.22: 50578958-50582999 on Build GRCh38
Transcripts
NM_005198.4 NR_027928.2

Gene Symbol
CHKB-CPT1B
Entrez Gene ID
386593
Gene Name
CHKB-CPT1B readthrough (NMD candidate)
Gene Aliases
CHKL-CPT1B
UniGene
Hs.439777
Gene Chromosome Location
Chr.22: 50568861-50582999 on Build GRCh38
Transcripts
NM_005198.4 NR_027928.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6497754 c.310C>T p.R104W Chr.22: 50582272-50582272 on Build GRCh38
3668638 c.316T>G p.Y106D Chr.22: 50582266-50582266 on Build GRCh38
4842563 c.298G>A p.E100K Chr.22: 50582284-50582284 on Build GRCh38
5779084 c.297G>A p.R99R Chr.22: 50582285-50582285 on Build GRCh38
3668637 c.317A>G p.Y106C Chr.22: 50582265-50582265 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs932865412 Chr.22: 50582171-50582171 on Build GRCh38
rs86337 Chr.22: 50582239-50582239 on Build GRCh38
rs373168311 Chr.22: 50582155-50582155 on Build GRCh38
rs977531813 Chr.22: 50582135-50582136 on Build GRCh38
rs564786154 Chr.22: 50582305-50582305 on Build GRCh38
rs146163970 Chr.22: 50582319-50582319 on Build GRCh38
rs923090455 Chr.22: 50582297-50582297 on Build GRCh38
rs1036112409 Chr.22: 50582216-50582216 on Build GRCh38
rs765518636 Chr.22: 50582282-50582282 on Build GRCh38
rs138208848 Chr.22: 50582192-50582192 on Build GRCh38
rs532210178 Chr.22: 50582308-50582308 on Build GRCh38
rs771547711 Chr.22: 50582243-50582243 on Build GRCh38
rs766423227 Chr.22: 50582293-50582293 on Build GRCh38
rs750678988 Chr.22: 50582289-50582289 on Build GRCh38
rs757428570 Chr.22: 50582291-50582291 on Build GRCh38
rs543205547 Chr.22: 50582258-50582258 on Build GRCh38
rs748097708 Chr.22: 50582229-50582229 on Build GRCh38
rs1037339903 Chr.22: 50582331-50582331 on Build GRCh38
rs865952423 Chr.22: 50582299-50582299 on Build GRCh38
rs745454865 Chr.22: 50582240-50582240 on Build GRCh38
rs990126817 Chr.22: 50582209-50582209 on Build GRCh38
rs751630140 Chr.22: 50582295-50582295 on Build GRCh38
rs574732831 Chr.22: 50582167-50582167 on Build GRCh38
rs560829170 Chr.22: 50582206-50582206 on Build GRCh38
rs371960270 Chr.22: 50582275-50582275 on Build GRCh38
rs376846624 Chr.22: 50582303-50582303 on Build GRCh38
rs894412168 Chr.22: 50582244-50582244 on Build GRCh38
rs868464194 Chr.22: 50582272-50582272 on Build GRCh38
rs1025792290 Chr.22: 50582143-50582143 on Build GRCh38
rs1003521884 Chr.22: 50582180-50582180 on Build GRCh38
rs369633837 Chr.22: 50582267-50582267 on Build GRCh38
rs775052422 Chr.22: 50582245-50582245 on Build GRCh38
rs924682608 Chr.22: 50582149-50582149 on Build GRCh38
rs967408054 Chr.22: 50582259-50582259 on Build GRCh38
rs756101150 Chr.22: 50582231-50582231 on Build GRCh38
rs1051288311 Chr.22: 50582175-50582175 on Build GRCh38
rs757118700 Chr.22: 50582310-50582310 on Build GRCh38
rs547428711 Chr.22: 50582320-50582320 on Build GRCh38
rs958954555 Chr.22: 50582196-50582196 on Build GRCh38
rs770798469 Chr.22: 50582235-50582235 on Build GRCh38
rs140867913 Chr.22: 50582307-50582307 on Build GRCh38
rs990109854 Chr.22: 50582152-50582152 on Build GRCh38
rs957397761 Chr.22: 50582150-50582150 on Build GRCh38

View Full Product Details