Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42752722-42752965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTBK2
Entrez Gene ID
146057
Gene Name
tau tubulin kinase 2
Gene Aliases
SCA11, TTBK
UniGene
Hs.646511
Gene Chromosome Location
Chr.15: 42738974-42920995 on Build GRCh38
Transcripts
NM_173500.3 XM_005254171.4 XM_005254173.4 XM_006720402.3 XM_006720403.3 XM_017021950.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5015283 c.2414T>C p.L805P Chr.15: 42752832-42752832 on Build GRCh38
7426227 c.2428G>A p.V810I Chr.15: 42752818-42752818 on Build GRCh38
324040 c.2519A>G p.N840S Chr.15: 42752727-42752727 on Build GRCh38
6567952 c.2311G>A p.E771K Chr.15: 42752935-42752935 on Build GRCh38
4846942 c.2335G>C p.E779Q Chr.15: 42752911-42752911 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377247038 Chr.15: 42752800-42752800 on Build GRCh38
rs375625245 Chr.15: 42752855-42752855 on Build GRCh38
rs1000137687 Chr.15: 42752844-42752844 on Build GRCh38
rs772736818 Chr.15: 42752882-42752882 on Build GRCh38
rs746184898 Chr.15: 42752865-42752865 on Build GRCh38
rs924397428 Chr.15: 42752890-42752890 on Build GRCh38
rs752741012 Chr.15: 42752753-42752753 on Build GRCh38
rs567793736 Chr.15: 42752762-42752762 on Build GRCh38
rs751291490 Chr.15: 42752798-42752798 on Build GRCh38
rs760713520 Chr.15: 42752887-42752887 on Build GRCh38
rs764259278 Chr.15: 42752760-42752760 on Build GRCh38
rs760767810 Chr.15: 42752757-42752757 on Build GRCh38
rs1044868862 Chr.15: 42752823-42752823 on Build GRCh38
rs749726502 Chr.15: 42752902-42752902 on Build GRCh38
rs544778041 Chr.15: 42752921-42752921 on Build GRCh38
rs748937770 Chr.15: 42752819-42752819 on Build GRCh38
rs777340522 Chr.15: 42752818-42752818 on Build GRCh38
rs778174365 Chr.15: 42752850-42752850 on Build GRCh38
rs776034291 Chr.15: 42752885-42752885 on Build GRCh38
rs200037227 Chr.15: 42752772-42752772 on Build GRCh38
rs758244281 Chr.15: 42752767-42752767 on Build GRCh38
rs373108991 Chr.15: 42752779-42752779 on Build GRCh38
rs766154799 Chr.15: 42752771-42752771 on Build GRCh38
rs1052988248 Chr.15: 42752893-42752893 on Build GRCh38
rs776781931 Chr.15: 42752930-42752930 on Build GRCh38
rs185441376 Chr.15: 42752828-42752828 on Build GRCh38
rs370373868 Chr.15: 42752814-42752814 on Build GRCh38
rs764217797 Chr.15: 42752900-42752900 on Build GRCh38
rs966127980 Chr.15: 42752848-42752848 on Build GRCh38
rs774867670 Chr.15: 42752860-42752860 on Build GRCh38
rs781138681 Chr.15: 42752812-42752812 on Build GRCh38
rs948795784 Chr.15: 42752751-42752751 on Build GRCh38

View Full Product Details