Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 18653359-18653626 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKL5
Entrez Gene ID
6792
Gene Name
cyclin dependent kinase like 5
Gene Aliases
CFAP247, EIEE2, ISSX, STK9
UniGene
Hs.659851
Gene Chromosome Location
Chr.X: 18425605-18653629 on Build GRCh38
Transcripts
NM_001037343.1 NM_003159.2 NM_000330.3

Gene Symbol
RS1
Entrez Gene ID
6247
Gene Name
retinoschisin 1
Gene Aliases
RS, XLRS1
UniGene
Hs.715725
Gene Chromosome Location
Chr.X: 18639688-18672103 on Build GRCh38
Transcripts
NM_001037343.1 NM_003159.2 NM_000330.3

SNP Detail

SNP ID Chromosome Location
rs58669795 Chr.X: 18653410-18653410 on Build GRCh38
rs1018524335 Chr.X: 18653480-18653480 on Build GRCh38
rs375094737 Chr.X: 18653385-18653385 on Build GRCh38
rs775556155 Chr.X: 18653433-18653433 on Build GRCh38
rs187317325 Chr.X: 18653548-18653548 on Build GRCh38
rs377160785 Chr.X: 18653494-18653494 on Build GRCh38
rs762576315 Chr.X: 18653534-18653534 on Build GRCh38
rs987192406 Chr.X: 18653455-18653455 on Build GRCh38
rs764866730 Chr.X: 18653594-18653594 on Build GRCh38
rs772980434 Chr.X: 18653518-18653518 on Build GRCh38
rs1047304798 Chr.X: 18653567-18653567 on Build GRCh38
rs910286513 Chr.X: 18653502-18653502 on Build GRCh38
rs587783162 Chr.X: 18653459-18653459 on Build GRCh38
rs369123557 Chr.X: 18653391-18653391 on Build GRCh38
rs587783163 Chr.X: 18653493-18653493 on Build GRCh38
rs139155110 Chr.X: 18653535-18653535 on Build GRCh38
rs767903007 Chr.X: 18653460-18653460 on Build GRCh38
rs267608666 Chr.X: 18653435-18653435 on Build GRCh38
rs753447480 Chr.X: 18653488-18653488 on Build GRCh38
rs35693326 Chr.X: 18653446-18653446 on Build GRCh38
rs727503848 Chr.X: 18653482-18653482 on Build GRCh38
rs763292733 Chr.X: 18653448-18653448 on Build GRCh38
rs756705837 Chr.X: 18653394-18653394 on Build GRCh38
rs752253030 Chr.X: 18653396-18653397 on Build GRCh38
rs762222076 Chr.X: 18653588-18653588 on Build GRCh38
rs768289707 Chr.X: 18653525-18653525 on Build GRCh38
rs573588032 Chr.X: 18653468-18653468 on Build GRCh38
rs759463462 Chr.X: 18653555-18653555 on Build GRCh38
rs150900695 Chr.X: 18653445-18653445 on Build GRCh38
rs776601149 Chr.X: 18653529-18653529 on Build GRCh38
rs34166184 Chr.X: 18653519-18653519 on Build GRCh38
rs779429355 Chr.X: 18653408-18653408 on Build GRCh38
rs145462429 Chr.X: 18653464-18653464 on Build GRCh38
rs201765217 Chr.X: 18653536-18653536 on Build GRCh38
rs267608394 Chr.X: 18653559-18653559 on Build GRCh38
rs779791138 Chr.X: 18653514-18653514 on Build GRCh38
rs757302280 Chr.X: 18653395-18653395 on Build GRCh38
rs766531184 Chr.X: 18653450-18653450 on Build GRCh38
rs753958338 Chr.X: 18653384-18653384 on Build GRCh38
rs764540699 Chr.X: 18653483-18653483 on Build GRCh38
rs942884617 Chr.X: 18653526-18653526 on Build GRCh38
rs994142937 Chr.X: 18653380-18653380 on Build GRCh38
rs745370971 Chr.X: 18653501-18653501 on Build GRCh38
rs757994307 Chr.X: 18653510-18653510 on Build GRCh38
rs36022183 Chr.X: 18653454-18653454 on Build GRCh38
rs267608667 Chr.X: 18653463-18653463 on Build GRCh38
rs764547426 Chr.X: 18653573-18653573 on Build GRCh38
rs771880879 Chr.X: 18653413-18653413 on Build GRCh38
rs754179699 Chr.X: 18653574-18653574 on Build GRCh38
rs775808084 Chr.X: 18653560-18653560 on Build GRCh38
rs376960593 Chr.X: 18653452-18653452 on Build GRCh38

View Full Product Details