Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 53432190-53432445 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD17B10
Entrez Gene ID
3028
Gene Name
hydroxysteroid 17-beta dehydrogenase 10
Gene Aliases
17b-HSD10, ABAD, CAMR, DUPXp11.22, ERAB, HADH2, HCD2, MHBD, MRPP2, MRX17, MRX31, MRXS10, SCHAD, SDR5C1
UniGene
Hs.171280
Gene Chromosome Location
Chr.X: 53431258-53434376 on Build GRCh38
Transcripts
NM_001037811.2 NM_004493.2

SNP Detail

SNP ID Chromosome Location
rs782330394 Chr.X: 53432331-53432331 on Build GRCh38
rs781979081 Chr.X: 53432236-53432236 on Build GRCh38
rs782314873 Chr.X: 53432222-53432222 on Build GRCh38
rs794729644 Chr.X: 53432386-53432386 on Build GRCh38
rs782143488 Chr.X: 53432281-53432281 on Build GRCh38
rs782087883 Chr.X: 53432250-53432250 on Build GRCh38
rs782357172 Chr.X: 53432257-53432257 on Build GRCh38
rs781966115 Chr.X: 53432371-53432371 on Build GRCh38
rs782197000 Chr.X: 53432220-53432220 on Build GRCh38
rs781916823 Chr.X: 53432320-53432320 on Build GRCh38
rs782725629 Chr.X: 53432322-53432322 on Build GRCh38
rs782818492 Chr.X: 53432353-53432353 on Build GRCh38
rs781982912 Chr.X: 53432265-53432265 on Build GRCh38
rs371014686 Chr.X: 53432345-53432345 on Build GRCh38
rs965610533 Chr.X: 53432318-53432318 on Build GRCh38
rs781891910 Chr.X: 53432365-53432365 on Build GRCh38
rs374438347 Chr.X: 53432381-53432381 on Build GRCh38
rs782466620 Chr.X: 53432409-53432409 on Build GRCh38
rs782705686 Chr.X: 53432300-53432300 on Build GRCh38
rs201378370 Chr.X: 53432321-53432321 on Build GRCh38
rs587777651 Chr.X: 53432347-53432347 on Build GRCh38
rs104886492 Chr.X: 53432410-53432410 on Build GRCh38

View Full Product Details