Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278511.1 | NP_001265440.1 | 7 - 8 | 1162 | 85 |
NM_001278512.1 | NP_001265441.1 | 8 - 9 | 1258 | 85 | |
NM_004644.4 | NP_004635.2 | 8 - 9 | 1258 | 85 | |
XM_017022640.1 | XP_016878129.1 | 8 - 9 | 1462 | 85 | |
XM_017022641.1 | XP_016878130.1 | 8 - 9 | 1462 | 85 | |
GenBank mRNA | AF022152.1 | - | 8 - 9 | 1087 | 85 |
BC093739.1 | - | 8 - 9 | 1129 | 85 | |
BC143342.1 | - | 7 - 8 | 1033 | 85 | |
BC143346.1 | - | 8 - 9 | 1129 | 85 | |
CR749860.1 | - | 7 - 8 | 1258 | 85 |