Assay Design



Gene Transcripts

Gene Symbol
ESPNL
Entrez Gene ID
339768
Gene Name
espin-like
Gene Aliases
-
Chromosome Location
Chr.2: 238100310 - 238133287 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194312.3 NP_919288.2 4 - 5 966 134
GenBank mRNA AK124559.1 - 4 - 5 891 134

View Full Product Details