Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 101031923-101032182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MUC17
Entrez Gene ID
140453
Gene Name
mucin 17, cell surface associated
Gene Aliases
MUC-17, MUC-3, MUC3
UniGene
Hs.271819
Gene Chromosome Location
Chr.7: 101020083-101058859 on Build GRCh38
Transcripts
NM_001040105.1 NR_133665.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3631152 c.656C>A p.T219K Chr.7: 101032072-101032072 on Build GRCh38
5543152 c.510C>T p.P170P Chr.7: 101031926-101031926 on Build GRCh38
7268844 c.751G>A p.A251T Chr.7: 101032167-101032167 on Build GRCh38
6275305 c.685G>T p.A229S Chr.7: 101032101-101032101 on Build GRCh38
237779 c.621C>G p.P207P Chr.7: 101032037-101032037 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754012525 Chr.7: 101031989-101031989 on Build GRCh38
rs748941063 Chr.7: 101032134-101032134 on Build GRCh38
rs760938068 Chr.7: 101032054-101032054 on Build GRCh38
rs774025440 Chr.7: 101032073-101032073 on Build GRCh38
rs747742194 Chr.7: 101032033-101032033 on Build GRCh38
rs761144835 Chr.7: 101032158-101032158 on Build GRCh38
rs758292037 Chr.7: 101032012-101032012 on Build GRCh38
rs143067910 Chr.7: 101032094-101032094 on Build GRCh38
rs772878703 Chr.7: 101032041-101032041 on Build GRCh38
rs147023140 Chr.7: 101032003-101032003 on Build GRCh38
rs372923916 Chr.7: 101031946-101031946 on Build GRCh38
rs762016528 Chr.7: 101032076-101032076 on Build GRCh38
rs762540897 Chr.7: 101032042-101032042 on Build GRCh38
rs765359937 Chr.7: 101032084-101032084 on Build GRCh38
rs748910623 Chr.7: 101032109-101032109 on Build GRCh38
rs927244054 Chr.7: 101031993-101031993 on Build GRCh38
rs201467055 Chr.7: 101032112-101032112 on Build GRCh38
rs116801454 Chr.7: 101032086-101032086 on Build GRCh38
rs556547867 Chr.7: 101032016-101032016 on Build GRCh38
rs865783682 Chr.7: 101031985-101031985 on Build GRCh38
rs777204968 Chr.7: 101032120-101032120 on Build GRCh38
rs746923995 Chr.7: 101032015-101032015 on Build GRCh38
rs868095285 Chr.7: 101032023-101032023 on Build GRCh38
rs560663109 Chr.7: 101032070-101032070 on Build GRCh38
rs756322676 Chr.7: 101031968-101031968 on Build GRCh38
rs750634422 Chr.7: 101032010-101032010 on Build GRCh38
rs779939140 Chr.7: 101032013-101032013 on Build GRCh38
rs753164036 Chr.7: 101031957-101031957 on Build GRCh38
rs770631238 Chr.7: 101032137-101032137 on Build GRCh38
rs143664405 Chr.7: 101032093-101032093 on Build GRCh38
rs764689052 Chr.7: 101031959-101031959 on Build GRCh38
rs763686003 Chr.7: 101032071-101032071 on Build GRCh38
rs371729341 Chr.7: 101032044-101032044 on Build GRCh38
rs540080829 Chr.7: 101032138-101032138 on Build GRCh38
rs779318632 Chr.7: 101032008-101032008 on Build GRCh38
rs10229731 Chr.7: 101032095-101032095 on Build GRCh38
rs905712151 Chr.7: 101032126-101032126 on Build GRCh38
rs775993341 Chr.7: 101032051-101032051 on Build GRCh38
rs141443818 Chr.7: 101031970-101031970 on Build GRCh38
rs769312461 Chr.7: 101032104-101032104 on Build GRCh38
rs748035347 Chr.7: 101032102-101032102 on Build GRCh38
rs375655258 Chr.7: 101031961-101031961 on Build GRCh38
rs780978510 Chr.7: 101032025-101032025 on Build GRCh38
rs890060564 Chr.7: 101032151-101032151 on Build GRCh38
rs769259358 Chr.7: 101032039-101032039 on Build GRCh38
rs761181335 Chr.7: 101031963-101031963 on Build GRCh38
rs781229156 Chr.7: 101032101-101032101 on Build GRCh38

View Full Product Details