Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 151027397-151027648 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPX3
Entrez Gene ID
2878
Gene Name
glutathione peroxidase 3
Gene Aliases
GPx-P, GSHPx-3, GSHPx-P
UniGene
Hs.386793
Gene Chromosome Location
Chr.5: 151020438-151028993 on Build GRCh38
Transcripts
NM_002084.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1064485 c.451T>G p.F151V Chr.5: 151027523-151027523 on Build GRCh38
1310863 c.406G>C p.E136Q Chr.5: 151027478-151027478 on Build GRCh38
4447856 c.424G>A p.G142R Chr.5: 151027496-151027496 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765755875 Chr.5: 151027574-151027574 on Build GRCh38
rs760257655 Chr.5: 151027524-151027524 on Build GRCh38
rs370194625 Chr.5: 151027486-151027486 on Build GRCh38
rs771492353 Chr.5: 151027434-151027434 on Build GRCh38
rs8177445 Chr.5: 151027454-151027454 on Build GRCh38
rs896941157 Chr.5: 151027619-151027619 on Build GRCh38
rs765489402 Chr.5: 151027457-151027457 on Build GRCh38
rs767581466 Chr.5: 151027442-151027442 on Build GRCh38
rs79510492 Chr.5: 151027533-151027533 on Build GRCh38
rs762300821 Chr.5: 151027453-151027453 on Build GRCh38
rs745591470 Chr.5: 151027516-151027516 on Build GRCh38
rs1007155425 Chr.5: 151027464-151027464 on Build GRCh38
rs771583068 Chr.5: 151027519-151027519 on Build GRCh38
rs373809425 Chr.5: 151027520-151027520 on Build GRCh38
rs76342174 Chr.5: 151027532-151027532 on Build GRCh38
rs758975709 Chr.5: 151027433-151027433 on Build GRCh38
rs558592006 Chr.5: 151027505-151027505 on Build GRCh38
rs909726764 Chr.5: 151027521-151027521 on Build GRCh38
rs754105125 Chr.5: 151027452-151027452 on Build GRCh38
rs371422003 Chr.5: 151027444-151027444 on Build GRCh38
rs750687719 Chr.5: 151027471-151027471 on Build GRCh38
rs777283698 Chr.5: 151027546-151027546 on Build GRCh38
rs201249129 Chr.5: 151027476-151027476 on Build GRCh38
rs776064760 Chr.5: 151027439-151027439 on Build GRCh38
rs1033602519 Chr.5: 151027429-151027429 on Build GRCh38
rs563091688 Chr.5: 151027595-151027595 on Build GRCh38
rs749039304 Chr.5: 151027495-151027495 on Build GRCh38
rs544404247 Chr.5: 151027593-151027593 on Build GRCh38
rs572854749 Chr.5: 151027440-151027440 on Build GRCh38
rs929716376 Chr.5: 151027626-151027626 on Build GRCh38
rs916465080 Chr.5: 151027598-151027598 on Build GRCh38
rs373449699 Chr.5: 151027472-151027472 on Build GRCh38
rs62382298 Chr.5: 151027563-151027563 on Build GRCh38
rs762212554 Chr.5: 151027556-151027556 on Build GRCh38
rs771262485 Chr.5: 151027532-151027533 on Build GRCh38
rs11544082 Chr.5: 151027502-151027502 on Build GRCh38
rs921170460 Chr.5: 151027592-151027592 on Build GRCh38
rs576911208 Chr.5: 151027535-151027535 on Build GRCh38
rs975762179 Chr.5: 151027558-151027558 on Build GRCh38
rs1046477779 Chr.5: 151027612-151027612 on Build GRCh38
rs77902022 Chr.5: 151027431-151027431 on Build GRCh38
rs949109235 Chr.5: 151027611-151027611 on Build GRCh38
rs777786115 Chr.5: 151027492-151027492 on Build GRCh38
rs977141962 Chr.5: 151027499-151027499 on Build GRCh38
rs752647532 Chr.5: 151027484-151027484 on Build GRCh38
rs540252597 Chr.5: 151027458-151027458 on Build GRCh38

View Full Product Details