Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 9739412-9739677 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR143
Entrez Gene ID
4935
Gene Name
G protein-coupled receptor 143
Gene Aliases
NYS6, OA1
UniGene
Hs.74124
Gene Chromosome Location
Chr.X: 9725413-9765965 on Build GRCh38
Transcripts
NM_000273.2 XM_005274541.3

SNP Detail

SNP ID Chromosome Location
rs778572122 Chr.X: 9739510-9739510 on Build GRCh38
rs906015045 Chr.X: 9739509-9739509 on Build GRCh38
rs752023852 Chr.X: 9739439-9739439 on Build GRCh38
rs756728406 Chr.X: 9739582-9739582 on Build GRCh38
rs140873054 Chr.X: 9739526-9739526 on Build GRCh38
rs151226761 Chr.X: 9739481-9739481 on Build GRCh38
rs748861491 Chr.X: 9739581-9739581 on Build GRCh38
rs748951376 Chr.X: 9739472-9739472 on Build GRCh38
rs745458128 Chr.X: 9739633-9739633 on Build GRCh38
rs771056170 Chr.X: 9739574-9739574 on Build GRCh38
rs776550135 Chr.X: 9739600-9739600 on Build GRCh38
rs759315924 Chr.X: 9739655-9739655 on Build GRCh38
rs138809166 Chr.X: 9739560-9739560 on Build GRCh38
rs755297765 Chr.X: 9739446-9739446 on Build GRCh38
rs1023047490 Chr.X: 9739572-9739572 on Build GRCh38
rs749933516 Chr.X: 9739475-9739476 on Build GRCh38
rs946950817 Chr.X: 9739451-9739451 on Build GRCh38
rs781743213 Chr.X: 9739447-9739447 on Build GRCh38
rs771928397 Chr.X: 9739654-9739654 on Build GRCh38
rs777471473 Chr.X: 9739550-9739550 on Build GRCh38
rs751704023 Chr.X: 9739554-9739554 on Build GRCh38
rs763113825 Chr.X: 9739546-9739546 on Build GRCh38
rs190174009 Chr.X: 9739469-9739469 on Build GRCh38
rs756548263 Chr.X: 9739571-9739571 on Build GRCh38
rs745715129 Chr.X: 9739497-9739497 on Build GRCh38
rs150179962 Chr.X: 9739527-9739527 on Build GRCh38
rs374388815 Chr.X: 9739530-9739530 on Build GRCh38
rs773870720 Chr.X: 9739492-9739492 on Build GRCh38
rs748581801 Chr.X: 9739452-9739452 on Build GRCh38
rs34324958 Chr.X: 9739569-9739569 on Build GRCh38
rs947549747 Chr.X: 9739474-9739474 on Build GRCh38
rs954542195 Chr.X: 9739593-9739593 on Build GRCh38
rs368826221 Chr.X: 9739470-9739470 on Build GRCh38
rs773371296 Chr.X: 9739545-9739545 on Build GRCh38
rs755421333 Chr.X: 9739555-9739555 on Build GRCh38
rs975131612 Chr.X: 9739443-9739443 on Build GRCh38

View Full Product Details