Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004623.4 | NP_004614.3 | 9 - 10 | 1146 | 164 |
GenBank mRNA | AF063602.1 | - | 9 - 10 | 1108 | 164 |
AF073887.1 | - | 9 - 10 | 1091 | 164 | |
AK090964.1 | - | 9 - 10 | 1226 | 164 | |
AK222562.1 | - | 9 - 10 | 1099 | 164 | |
AK292426.1 | - | 9 - 10 | 1107 | 164 | |
BC001276.1 | - | 9 - 10 | 1085 | 164 | |
BT006817.1 | - | 9 - 10 | 1059 | 164 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_037639.2 | 32 - 33 | 5237 | 164 | |
NR_037640.2 | 32 - 33 | 5104 | 164 | ||
NR_037641.2 | 32 - 33 | 4883 | 164 | ||
GenBank mRNA | AK127008.1 | - | 16 - 17 | 2738 | 164 |
BX648088.1 | - | 32 - 33 | 5043 | 164 | |
CR749830.1 | - | 32 - 33 | 5189 | 164 |