Assay Design



Gene Transcripts

Gene Symbol
TAS2R45
Entrez Gene ID
259291
Gene Name
taste 2 receptor member 45
Gene Aliases
GPR59, T2R45, ZG24P
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176886.2 NP_795367.2 3 - 3 546 114
GenBank mRNA BC101154.1 - 3 - 3 546 114
BC101156.1 - 3 - 3 546 114

Gene Symbol
TAS2R43
Entrez Gene ID
259289
Gene Name
taste 2 receptor member 43
Gene Aliases
T2R43, T2R52
Chromosome Location
Chr.12: 11091287 - 11092313 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC101155.1 - 3 - 3 546 114

View Full Product Details