Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243342.1 | NP_001230271.1 | 4 - 5 | 708 | 90 |
NM_017950.3 | NP_060420.2 | 4 - 5 | 708 | 90 | |
XM_005257492.4 | XP_005257549.1 | 4 - 5 | 708 | 90 | |
XM_011524963.2 | XP_011523265.1 | 4 - 5 | 745 | 90 | |
XM_011524965.2 | XP_011523267.1 | 4 - 5 | 708 | 90 | |
XM_017024807.1 | XP_016880296.1 | 4 - 5 | 708 | 90 | |
GenBank mRNA | AK000760.1 | - | 4 - 5 | 688 | 90 |
BC035251.1 | - | 4 - 5 | 701 | 90 | |
BC058288.1 | - | 4 - 5 | 686 | 90 |