Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143989.2 | NP_001137461.1 | 8 - 9 | 1187 | 91 |
XM_011540763.1 | XP_011539065.1 | 9 - 10 | 1260 | 91 | |
GenBank mRNA | AK057395.1 | - | 6 - 7 | 796 | 91 |
AK302318.1 | - | 8 - 9 | 1182 | 91 | |
AK316438.1 | - | 8 - 9 | 1173 | 91 | |
BC125162.1 | - | 8 - 9 | 1148 | 91 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143987.1 | NP_001137459.1 | 9 - 10 | 1274 | 91 |
NM_001143988.1 | NP_001137460.1 | 8 - 9 | 1187 | 91 | |
XM_005271144.3 | XP_005271201.1 | 8 - 9 | 1311 | 91 | |
XM_011542012.2 | XP_011540314.1 | 13 - 14 | 2640 | 91 | |
XM_011542014.2 | XP_011540316.1 | 8 - 9 | 1949 | 91 | |
XM_017002148.1 | XP_016857637.1 | 13 - 14 | 1775 | 91 | |
GenBank mRNA | BC125161.1 | - | 9 - 10 | 1235 | 91 |