Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 63061327-63061564 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTPA
Entrez Gene ID
7274
Gene Name
alpha tocopherol transfer protein
Gene Aliases
ATTP, AVED, TTP1, alphaTTP
UniGene
Hs.69049
Gene Chromosome Location
Chr.8: 63058409-63086523 on Build GRCh38
Transcripts
NM_000370.3 XM_006716468.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3900938 c.715G>T p.D239Y Chr.8: 63061374-63061374 on Build GRCh38
1100991 c.700C>A p.L234I Chr.8: 63061389-63061389 on Build GRCh38
2720432 c.711C>T p.F237F Chr.8: 63061378-63061378 on Build GRCh38
2720433 c.695G>A p.S232N Chr.8: 63061394-63061394 on Build GRCh38
3834892 c.750C>G p.F250L Chr.8: 63061339-63061339 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762716411 Chr.8: 63061443-63061443 on Build GRCh38
rs779339516 Chr.8: 63061367-63061367 on Build GRCh38
rs752997771 Chr.8: 63061457-63061457 on Build GRCh38
rs374858226 Chr.8: 63061469-63061469 on Build GRCh38
rs577508179 Chr.8: 63061398-63061398 on Build GRCh38
rs746629810 Chr.8: 63061445-63061460 on Build GRCh38
rs559233106 Chr.8: 63061357-63061357 on Build GRCh38
rs776505121 Chr.8: 63061412-63061412 on Build GRCh38
rs767759228 Chr.8: 63061447-63061447 on Build GRCh38
rs397515526 Chr.8: 63061353-63061353 on Build GRCh38
rs2045224 Chr.8: 63061429-63061429 on Build GRCh38
rs370191429 Chr.8: 63061444-63061444 on Build GRCh38
rs760144623 Chr.8: 63061399-63061399 on Build GRCh38
rs34766023 Chr.8: 63061458-63061458 on Build GRCh38
rs35250281 Chr.8: 63061488-63061488 on Build GRCh38
rs771738459 Chr.8: 63061378-63061378 on Build GRCh38
rs764874169 Chr.8: 63061422-63061422 on Build GRCh38
rs535045937 Chr.8: 63061473-63061473 on Build GRCh38
rs537647819 Chr.8: 63061351-63061351 on Build GRCh38
rs374388749 Chr.8: 63061376-63061376 on Build GRCh38
rs771362888 Chr.8: 63061359-63061359 on Build GRCh38
rs761757308 Chr.8: 63061421-63061421 on Build GRCh38
rs777427548 Chr.8: 63061436-63061437 on Build GRCh38
rs768187989 Chr.8: 63061411-63061411 on Build GRCh38
rs1027551217 Chr.8: 63061498-63061498 on Build GRCh38

View Full Product Details