Assay Design



Gene Transcripts

Gene Symbol
AMIGO1
Entrez Gene ID
57463
Gene Name
adhesion molecule with Ig like domain 1
Gene Aliases
ALI2, AMIGO, AMIGO-1
Chromosome Location
Chr.1: 109504178 - 109509742 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020703.2 NP_065754.2 1 - 2 296 70
GenBank mRNA AK295678.1 - 1 - 2 275 70
BC040879.1 - 1 - 2 264 70
DC317707.1 - 1 - 2 298 70

View Full Product Details