Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 49186653-49186902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOLH1
Entrez Gene ID
2346
Gene Name
folate hydrolase (prostate-specific membrane antigen) 1
Gene Aliases
FGCP, FOLH, GCP2, GCPII, NAALAD1, NAALAdase, PSM, PSMA, mGCP
UniGene
Hs.645352
Gene Chromosome Location
Chr.11: 49145094-49208706 on Build GRCh38
Transcripts
NM_001014986.1 NM_001193471.1 NM_001193472.1 NM_001193473.1 NM_004476.1 XM_011519958.2 XM_017017432.1 XM_017017433.1 XM_017017434.1 XM_017017435.1 XM_017017436.1 XM_017017437.1 XM_017017438.1 XM_017017439.1 XM_017017440.1 XM_017017441.1 XM_017017442.1 XM_017017443.1 XM_017017444.1 XM_017017445.1 XM_017017446.1 XM_017017447.1 XM_017017448.1 XM_017017449.1 XM_017017450.1 XM_017017451.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4033410 c.530T>G p.V177G Chr.11: 49186753-49186753 on Build GRCh38
221545 c.584A>G p.N195S Chr.11: 49186699-49186699 on Build GRCh38
320480 c.630A>T p.R210S Chr.11: 49186653-49186653 on Build GRCh38
6665982 c.526T>C p.Y176H Chr.11: 49186757-49186757 on Build GRCh38
5050864 c.516C>T p.G172G Chr.11: 49186767-49186767 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs75655145 Chr.11: 49186746-49186746 on Build GRCh38
rs568557023 Chr.11: 49186780-49186780 on Build GRCh38
rs767819649 Chr.11: 49186862-49186862 on Build GRCh38
rs776299190 Chr.11: 49186718-49186718 on Build GRCh38
rs757958518 Chr.11: 49186741-49186741 on Build GRCh38
rs377095633 Chr.11: 49186742-49186742 on Build GRCh38
rs778134535 Chr.11: 49186766-49186766 on Build GRCh38
rs768761870 Chr.11: 49186800-49186800 on Build GRCh38
rs773252374 Chr.11: 49186813-49186813 on Build GRCh38
rs74858499 Chr.11: 49186781-49186781 on Build GRCh38
rs772191107 Chr.11: 49186705-49186705 on Build GRCh38
rs747193293 Chr.11: 49186785-49186785 on Build GRCh38
rs138028811 Chr.11: 49186714-49186714 on Build GRCh38
rs761741517 Chr.11: 49186719-49186719 on Build GRCh38
rs769759209 Chr.11: 49186806-49186806 on Build GRCh38
rs989860615 Chr.11: 49186850-49186850 on Build GRCh38
rs750442389 Chr.11: 49186737-49186737 on Build GRCh38
rs77530815 Chr.11: 49186881-49186881 on Build GRCh38
rs745938739 Chr.11: 49186681-49186681 on Build GRCh38
rs531736406 Chr.11: 49186856-49186856 on Build GRCh38
rs75111588 Chr.11: 49186715-49186715 on Build GRCh38
rs754599970 Chr.11: 49186761-49186761 on Build GRCh38
rs981498774 Chr.11: 49186825-49186825 on Build GRCh38
rs776975754 Chr.11: 49186804-49186804 on Build GRCh38
rs79155991 Chr.11: 49186767-49186767 on Build GRCh38
rs755366873 Chr.11: 49186844-49186844 on Build GRCh38
rs377049265 Chr.11: 49186777-49186777 on Build GRCh38
rs775565658 Chr.11: 49186782-49186782 on Build GRCh38
rs775663569 Chr.11: 49186711-49186711 on Build GRCh38
rs765257997 Chr.11: 49186732-49186732 on Build GRCh38
rs945875198 Chr.11: 49186877-49186877 on Build GRCh38
rs61886509 Chr.11: 49186827-49186827 on Build GRCh38

View Full Product Details