Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004559.4 | NP_004550.2 | 8 - 8 | 1559 | 160 |
NR_132737.1 | 7 - 7 | 1377 | 160 | ||
GenBank mRNA | BC002411.2 | - | 8 - 8 | 1342 | 160 |
BC010430.1 | - | 8 - 8 | 1340 | 160 | |
BC013838.1 | - | 7 - 7 | 1377 | 160 | |
BC015208.1 | - | 8 - 8 | 1291 | 160 | |
BC020164.1 | - | 1 - 1 | 800 | 160 | |
BC038384.1 | - | 8 - 8 | 1348 | 160 | |
BC065571.1 | - | 8 - 8 | 1366 | 160 | |
BC070084.1 | - | 8 - 8 | 1366 | 160 | |
BC071708.1 | - | 8 - 8 | 1350 | 160 | |
BC090038.1 | - | 8 - 8 | 1351 | 160 | |
BC106045.1 | - | 8 - 8 | 1353 | 160 | |
L28809.1 | - | 8 - 8 | 1309 | 160 |