Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015155.2 | NP_055970.1 | 18 - 18 | 5765 | 120 |
XM_017015987.1 | XP_016871476.1 | 17 - 17 | 7919 | 120 | |
XM_017015988.1 | XP_016871477.1 | 17 - 17 | 7812 | 120 | |
XM_017015989.1 | XP_016871478.1 | 17 - 17 | 7857 | 120 | |
XM_017015992.1 | XP_016871481.1 | 13 - 13 | 5133 | 120 | |
XM_017015993.1 | XP_016871482.1 | 16 - 16 | 5519 | 120 | |
XM_017015994.1 | XP_016871483.1 | 17 - 17 | 5451 | 120 | |
GenBank mRNA | BC003381.1 | - | 1 - 1 | 422 | 120 |
BC152443.1 | - | 17 - 17 | 5489 | 120 | |
D86971.2 | - | 17 - 17 | 5489 | 120 |