Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 53433673-53433934 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD17B10
Entrez Gene ID
3028
Gene Name
hydroxysteroid 17-beta dehydrogenase 10
Gene Aliases
17b-HSD10, ABAD, CAMR, DUPXp11.22, ERAB, HADH2, HCD2, MHBD, MRPP2, MRX17, MRX31, MRXS10, SCHAD, SDR5C1
UniGene
Hs.171280
Gene Chromosome Location
Chr.X: 53431258-53434376 on Build GRCh38
Transcripts
NM_001037811.2 NM_004493.2

SNP Detail

SNP ID Chromosome Location
rs782502153 Chr.X: 53433788-53433788 on Build GRCh38
rs200274641 Chr.X: 53433817-53433817 on Build GRCh38
rs371890650 Chr.X: 53433858-53433858 on Build GRCh38
rs782197997 Chr.X: 53433856-53433856 on Build GRCh38
rs945465107 Chr.X: 53433737-53433737 on Build GRCh38
rs781980924 Chr.X: 53433903-53433903 on Build GRCh38
rs782608162 Chr.X: 53433866-53433866 on Build GRCh38
rs781906672 Chr.X: 53433783-53433783 on Build GRCh38
rs12847788 Chr.X: 53433879-53433879 on Build GRCh38
rs782748623 Chr.X: 53433816-53433816 on Build GRCh38
rs781946533 Chr.X: 53433708-53433708 on Build GRCh38
rs376193455 Chr.X: 53433755-53433755 on Build GRCh38
rs782381265 Chr.X: 53433893-53433893 on Build GRCh38
rs782719811 Chr.X: 53433773-53433773 on Build GRCh38
rs367982176 Chr.X: 53433767-53433767 on Build GRCh38
rs782123415 Chr.X: 53433726-53433726 on Build GRCh38
rs368358217 Chr.X: 53433703-53433703 on Build GRCh38
rs782029755 Chr.X: 53433769-53433769 on Build GRCh38
rs978092932 Chr.X: 53433768-53433768 on Build GRCh38
rs782158864 Chr.X: 53433770-53433770 on Build GRCh38
rs782685393 Chr.X: 53433855-53433855 on Build GRCh38
rs782505983 Chr.X: 53433824-53433824 on Build GRCh38
rs781794376 Chr.X: 53433779-53433779 on Build GRCh38
rs140876830 Chr.X: 53433740-53433740 on Build GRCh38
rs782283907 Chr.X: 53433914-53433914 on Build GRCh38
rs782206781 Chr.X: 53433877-53433877 on Build GRCh38
rs781871620 Chr.X: 53433820-53433820 on Build GRCh38

View Full Product Details