Assay Design
Gene Transcripts
- Gene Symbol
- AK6
- Entrez Gene ID
-
102157402
- Gene Name
- adenylate kinase 6
- Gene Aliases
-
AD-004, CGI-137, CINAP, CIP, hCINAP
- Chromosome Location
-
Chr.5: 69351726 - 69370013 on Build GRCh38
- Gene Symbol
- TAF9
- Entrez Gene ID
-
6880
- Gene Name
- TATA-box binding protein associated factor 9
- Gene Aliases
-
MGC:5067, STAF31/32, TAF2G, TAFII-31, TAFII-32, TAFII31, TAFII32, TAFIID32
- Chromosome Location
-
Chr.5: 69364743 - 69370013 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC007349.2 |
- |
5 - 5 |
575 |
149 |
View Full Product Details