Assay Design



Gene Transcripts

Gene Symbol
PRSS57
Entrez Gene ID
400668
Gene Name
protease, serine 57
Gene Aliases
NSP4, PRSSL1, UNQ782
Chromosome Location
Chr.19: 685521 - 695461 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308209.1 NP_001295138.1 4 - 5 674 107
NM_214710.4 NP_999875.1 4 - 5 677 107
GenBank mRNA AY358594.1 - 4 - 5 714 107
BC137161.1 - 4 - 5 679 107
BC144505.1 - 4 - 5 676 107

View Full Product Details