Assay Design



Gene Transcripts

Gene Symbol
SERINC4
Entrez Gene ID
619189
Gene Name
serine incorporator 4
Gene Aliases
-
Chromosome Location
Chr.15: 43794174 - 43800097 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258031.1 NP_001244960.1 5 - 6 745 67
GenBank mRNA AK097682.1 - 3 - 4 351 67

View Full Product Details