Assay Design



Gene Transcripts

Gene Symbol
TCEB2
Entrez Gene ID
6923
Gene Name
transcription elongation factor B subunit 2
Gene Aliases
ELOB, SIII
Chromosome Location
Chr.16: 2771414 - 2777296 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007108.3 NP_009039.1 3 - 4 302 62
NM_207013.2 NP_996896.1 3 - 4 302 62
GenBank mRNA BC013306.2 - 2 - 3 256 62
BC065000.1 - 3 - 4 258 62
BF967495.2 - 3 - 4 308 62
BG329435.1 - - 283 62
BM553320.1 - 3 - 4 317 62
BM700019.1 - 2 - 3 253 62
BM921799.1 - 2 - 3 270 62
BU533433.1 - 3 - 4 273 62
L42856.1 - 2 - 3 245 62

View Full Product Details