Product Details

Assay Reference Genome
Location

Chr.7:37896984 on build GRCh38
Cytoband
7p14.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
51314
Gene Symbol
NME8
Gene Name
NME/NM23 family member 8
Gene Aliases
CILD6, HEL-S-99, NM23-H8, SPTRX2, TXNDC3, sptrx-2
Location
Chr.7:37848597-37900401 on build GRCh38
Assay Gene Location
Within Exon 17
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
NME8 NM_016616.4 17 2031 NP_057700.3
AF202051.2 16 1760 AAF20909.2
AF305596.1 16 1729 AAN04258.1
AK313599.1 16 1771
BC036816.1 17 2031 AAH36816.1
GQ472221.1 17 2031

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv24163 Chr7:37891422 - 37897099 on Build GRCh38 Gain NME8
esv3612864 Chr7:37891337 - 37897492 on Build GRCh38 Gain NME8
nsv8074 Chr7:37825738 - 38170485 on Build GRCh38 Loss EPDR1 NME8 SFRP4

View Full Product Details