Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032538.1 | NP_115927.1 | 3 - 4 | 533 | 55 |
XM_011514947.2 | XP_011513249.1 | 2 - 3 | 323 | 55 | |
XM_017011364.1 | XP_016866853.1 | 2 - 3 | 323 | 55 | |
XM_017011365.1 | XP_016866854.1 | 2 - 3 | 323 | 55 | |
XM_017011366.1 | XP_016866855.1 | 2 - 3 | 323 | 55 | |
XM_017011367.1 | XP_016866856.1 | 2 - 3 | 323 | 55 | |
GenBank mRNA | AB058758.1 | - | 1 - 2 | 102 | 55 |
AB218664.1 | - | 4 - 5 | 337 | 55 | |
AK131217.1 | - | 2 - 3 | 263 | 55 |