Assay Design



Gene Transcripts

Gene Symbol
MUC5AC
Entrez Gene ID
4586
Gene Name
mucin 5AC, oligomeric mucus/gel-forming
Gene Aliases
MUC5, TBM, leB, mucin
Chromosome Location
Chr.11: 1157953 - 1201138 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001304359.1 NP_001291288.1 - 1291 78
GenBank mRNA AF015521.1 - 10 - 11 1291 78

View Full Product Details