Assay Design



Gene Transcripts

Gene Symbol
LACTBL1
Entrez Gene ID
646262
Gene Name
lactamase beta like 1
Gene Aliases
-
Chromosome Location
Chr.1: 22953043 - 22972316 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289974.1 NP_001276903.1 1 - 2 150 145

View Full Product Details