Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001669.2 | NP_001001669.2 | 6 - 7 | 855 | 84 |
XM_005268447.4 | XP_005268504.1 | 6 - 7 | 826 | 84 | |
XM_005268448.1 | XP_005268505.1 | 5 - 6 | 729 | 84 | |
XM_006714784.3 | XP_006714847.1 | 7 - 8 | 1194 | 84 | |
XM_011537642.2 | XP_011535944.1 | 6 - 7 | 877 | 84 | |
XM_017009460.1 | XP_016864949.1 | 6 - 7 | 1079 | 84 | |
XM_017009461.1 | XP_016864950.1 | 5 - 6 | 729 | 84 | |
GenBank mRNA | AB199796.1 | - | 5 - 6 | 802 | 84 |
AK300355.1 | - | 5 - 6 | 716 | 84 | |
AK300559.1 | - | 6 - 7 | 954 | 84 | |
AK316002.1 | - | 6 - 7 | 925 | 84 | |
BC041325.1 | - | 6 - 7 | 855 | 84 |