Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001669.2 | NP_001001669.2 | 8 - 9 | 1070 | 55 |
XM_005268447.4 | XP_005268504.1 | 8 - 9 | 1041 | 55 | |
XM_005268448.1 | XP_005268505.1 | 7 - 8 | 944 | 55 | |
XM_006714784.3 | XP_006714847.1 | 9 - 10 | 1409 | 55 | |
XM_011537642.2 | XP_011535944.1 | 8 - 9 | 1092 | 55 | |
GenBank mRNA | AB199796.1 | - | 7 - 8 | 1017 | 55 |
AK300355.1 | - | 7 - 8 | 931 | 55 | |
AK300559.1 | - | 8 - 9 | 1169 | 55 | |
AK316002.1 | - | 8 - 9 | 1140 | 55 | |
BC041325.1 | - | 8 - 9 | 1070 | 55 |