Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114726.2 | NP_001108198.2 | 2 - 3 | 852 | 88 |
NM_001174164.1 | NP_001167635.1 | 3 - 4 | 969 | 88 | |
XM_005250342.3 | XP_005250399.1 | 2 - 3 | 969 | 88 | |
XM_005250343.3 | XP_005250400.1 | 2 - 3 | 878 | 88 | |
XM_006715985.3 | XP_006716048.1 | 2 - 3 | 982 | 88 | |
XM_017012205.1 | XP_016867694.1 | 3 - 4 | 1788 | 88 | |
XM_017012206.1 | XP_016867695.1 | 2 - 3 | 1761 | 88 | |
GenBank mRNA | BX327107.2 | - | 1 - 2 | 134 | 88 |