Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000083.2 | NP_000074.2 | 12 - 13 | 1493 | 86 |
XM_011515782.2 | XP_011514084.1 | 2 - 3 | 241 | 86 | |
XM_017011739.1 | XP_016867228.1 | 9 - 10 | 1218 | 86 | |
XM_017011740.1 | XP_016867229.1 | 9 - 10 | 1194 | 86 | |
GenBank mRNA | BC112156.1 | - | 12 - 13 | 1458 | 86 |
BC113495.1 | - | 12 - 13 | 1458 | 86 | |
M97820.1 | - | 9 - 10 | 897 | 86 | |
Z25884.1 | - | 12 - 13 | 1493 | 86 |